Skip to Content
Merck
All Photos(1)

Key Documents

596485

Sigma-Aldrich

Benzotriazole-1-carboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H6N4O
CAS Number:
Molecular Weight:
162.15
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

form

solid

Quality Level

reaction suitability

reaction type: C-C Bond Formation

mp

170-175 °C (lit.)

functional group

amine

SMILES string

NC(=O)n1nnc2ccccc12

InChI

1S/C7H6N4O/c8-7(12)11-6-4-2-1-3-5(6)9-10-11/h1-4H,(H2,8,12)

InChI key

UUMXWUNNKCQWHS-UHFFFAOYSA-N

Application

Benzotriazole-1-carboxamide can be used:
  • As a corrosion inhibitor, applicable for copper in the HNO3 solution.
  • As a reagent for the preparation of urea derivatives with mono- and N, N-disubstitution products.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dry and wet lab studies for some benzotriazole derivatives as possible corrosion inhibitors for copper in 1.0 M HNO3
Khaled KF and Amin MA
Corrosion Science, 51(9), 2098-2106 (2009)
Synthesis of mono-and N, N-disubstituted ureas
Katritzky AR, et al.
ARKIVOC (Gainesville, FL, United States), 8, 8-14 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service