All Photos(2)
About This Item
Linear Formula:
CaI2
CAS Number:
Molecular Weight:
293.89
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99%
form:
powder and chunks
Recommended Products
Quality Level
Assay
99%
form
powder and chunks
mp
779 °C (lit.)
SMILES string
I[Ca]I
InChI
1S/Ca.2HI/h;2*1H/q+2;;/p-2
InChI key
UNMYWSMUMWPJLR-UHFFFAOYSA-L
Looking for similar products? Visit Product Comparison Guide
Related Categories
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service