Skip to Content
Merck
All Photos(1)

Documents

223808

Sigma-Aldrich

Barium iodide dihydrate

≥95%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Bal2 · 2H2O
CAS Number:
Molecular Weight:
427.17
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

Assay

≥95%

form

powder and chunks

density

5.15 g/mL at 25 °C (lit.)

SMILES string

[H]O[H].[H]O[H].I[Ba]I

InChI

1S/Ba.2HI.2H2O/h;2*1H;2*1H2/q+2;;;;/p-2

InChI key

LRUCFPBOVNFNMO-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service