Skip to Content
Merck
All Photos(1)

Documents

590517

Sigma-Aldrich

2-Bromo-N,N-dimethylaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H10BrN
CAS Number:
Molecular Weight:
200.08
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

SMILES string

CN(C)c1ccccc1Br

InChI

1S/C8H10BrN/c1-10(2)8-6-4-3-5-7(8)9/h3-6H,1-2H3

InChI key

ONMSBNJJCUCYED-UHFFFAOYSA-N

Application

2-Bromo-N,N-dimethylaniline may be used in the synthesis of the following:
  • bis[(2-dimethylamino)phenyl]amine via a multi-step reaction procedure
  • 2-(N,N-dimethylaminophenyl)-bis(diethylamino)phosphine
  • 6-[2-(dimethylamino)phenyl]-2,2-bipyridine obtained via lithiation followed by reaction with 2,2-bipyridine
  • 1-(2′-(dimethylamino)phenyl)-2-diphenylphosphino-3-methylimidazolium trifluoromethanesulfonate via a multi-step reaction procedure

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Aquatic Acute 1 - Aquatic Chronic 1 - Carc. 2 - Eye Irrit. 2 - STOT RE 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

?Synthesis, Structures, and Luminescent Properties of d10 Group 12 Metal Complexes with Substituted 2,2-Bipyridine Ligands?
Liu m-x, et al.
European Journal of Inorganic Chemistry, 2006(21), 4317-4323 (2006)
?Rhodium complexes with a new chiral nitrogencontaining
Binol-based diphosphite or phosphonite ligand: synthesis and application to hydroformylation of styrene and/or hydrogenation of prochiral olefins?
Kostas.D I
Applied Organometallic Chemistry, 19(10), 1090-1095 (2005)
?Synthesis, Structures, and Luminescent Properties of d10 Group 12 Metal Complexes with Substituted 2,2-Bipyridine Ligands?
Chen J-S, et al.
J. Mol. Catal. A: Chem., 396, 68-76 (2015)
?Pd, Pt, and Ru complexes of a pincer bis(amino)amide ligand?
Ren P, et al.
Dalton Transactions, 40(35), 8906-8911 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service