Skip to Content
Merck
All Photos(1)

Key Documents

559318

Sigma-Aldrich

1-(2-Bromoethyl)naphthalene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H11Br
CAS Number:
Molecular Weight:
235.12
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

refractive index

n20/D 1.638 (lit.)

bp

237-238 °C/760 mmHg (lit.)

density

1.646 g/mL at 25 °C (lit.)

SMILES string

BrCCc1cccc2ccccc12

InChI

1S/C12H11Br/c13-9-8-11-6-3-5-10-4-1-2-7-12(10)11/h1-7H,8-9H2

InChI key

GPHCPUFIWQJZOI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-(2-Bromoethyl)naphthalene can be obtained from the reaction between 1-(2-hydroxyethyl)naphthalene and phosphorus tribromide.

Application

1-(2-Bromoethyl)naphthalene may be used to synthesize 2-iodo-1-[2-(1-naphthyl)ethyl]-1H-benzimidazole.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potentially carcinogenic cyclopenta [a] phenanthrenes. Part IV. Synthesis of 17-ketones by the Stobbe condensation.
Coombs MM, et al.
J. Chem. Soc. Sect. C, 9, 1266-1271 (1970)
Access to aromatic ring-fused benzimidazoles using photochemical substitutions of the benzimidazol-2-yl radical.
O'Connell JM, et al.
Synthesis, 44(12), 3371-3377 (2012)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service