Skip to Content
Merck
All Photos(1)

Documents

552313

Sigma-Aldrich

4-Phenylbenzylamine

97%

Synonym(s):

(4-Phenylphenyl)methanamine, 1-(Biphenyl-4-yl)methanamine, 4-(Aminomethyl)biphenyl, 4-Biphenylmethylamine, Biphenyl-4-ylmethanamine, [(1,1′-Biphenyl-4-yl)methyl]amine, [(Biphenyl-4-yl)methyl]amine, [1,1′-Biphenyl]-4-ylmethanamine, p-Phenylbenzylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5C6H4CH2NH2
CAS Number:
Molecular Weight:
183.25
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

48-53 °C (lit.)

SMILES string

NCc1ccc(cc1)-c2ccccc2

InChI

1S/C13H13N/c14-10-11-6-8-13(9-7-11)12-4-2-1-3-5-12/h1-9H,10,14H2

InChI key

RMSPOVPGDBDYKH-UHFFFAOYSA-N

Related Categories

Application

4-Phenylbenzylamine may be used in the synthesis of:
  • squaric acid asymmetric diamides of glycopeptide antibiotics
  • asymmetric diamides of daunomycin and carminomycin
  • N-(biphenyl-4-ylmethyl)-1-hydroxy-6-thioxo-1,6-dihydropyridine-2-carboxamide

4-Phenylbenzylamine may also be used in the synthesis of Gd3+ chelates of DOTA(1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetate) type ligands.

Other Notes

Handle and store under nitrogen to limit formation of impurities.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

224.6 °F - closed cup

Flash Point(C)

107 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new class of semisynthetic anthracycline glycoside antibiotics incorporating a squaric acid moiety.
Sztaricskai F, et al.
The Journal of Antibiotics, 58(11), 704-704 (2005)
chemical society(Great Britain)
Journal of the Chemical Society, Part 1 (1993)
A new series of glycopeptide antibiotics incorporating a squaric acid moiety.
Sztaricskai F, et al.
The Journal of Antibiotics, 59(9), 564-564 (2006)
Coupling fast water exchange to slow molecular tumbling in Gd3+ chelates: why faster is not always better.
Avedano S, et al.
Inorganic Chemistry, 52(15), 8436-8450 (2013)
Thioamide hydroxypyrothiones supersede amide hydroxypyrothiones in potency against anthrax lethal factor.
Agrawal A, et al.
Journal of Medicinal Chemistry, 52(4), 1063-1074 (2009)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service