Skip to Content
Merck
All Photos(1)

Documents

A53605

Sigma-Aldrich

Benzhydrylamine

97%

Synonym(s):

α-Aminodiphenylmethane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2CHNH2
CAS Number:
Molecular Weight:
183.25
Beilstein:
776434
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.595 (lit.)

bp

295 °C (lit.)

mp

12 °C (lit.)

density

1.063 g/mL at 25 °C (lit.)

SMILES string

NC(c1ccccc1)c2ccccc2

InChI

1S/C13H13N/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10,13H,14H2

InChI key

MGHPNCMVUAKAIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abdul M Seayad et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 18(18), 5693-5700 (2012-03-23)
A robust heterogeneous self-supported chiral titanium cluster (SCTC) catalyst and its application in the enantioselective imine-cyanation/Strecker reaction is described under batch and continuous processes. One of the major hurdles in the asymmetric Strecker reaction is the lack of availability of
Adrenergic blocking drugs; blocking of excitatory responses to epinephrine and adrenergic nerve stimulation with N-alkyl-N-(2-chloroethyl)-benzhydrylamines.
E R LOEW et al.
The Journal of pharmacology and experimental therapeutics, 97(4), 441-449 (1949-12-01)
Fang-Kun Deng et al.
Journal of peptide science : an official publication of the European Peptide Society, 16(10), 545-550 (2010-09-09)
Well-characterized resins of high purity are critical for effective solid phase peptide synthesis (SPPS). The quality of commercial (4-methyl)benzhydrylamine-resin (MBHA-resin), used for the synthesis of peptide amides, is not consistent and residual ketone functionalities are frequently present. Such ketone or
Dianne Alewood et al.
Journal of peptide science : an official publication of the European Peptide Society, 16(10), 551-557 (2010-09-24)
The standard p-MBHA resin used during Boc-chemistry synthesis of peptides carrying C-terminal carboxamides is compromised by batch-to-batch variations in its performance. This can cause artificially 'difficult' couplings during peptide chain assembly, which may ultimately lead to failed syntheses given the
Regina S H Carvalho et al.
Journal of biochemical and biophysical methods, 57(1), 75-83 (2003-07-02)
Benzhydrylamine-resin (BHAR), a copolymer of styrene-divinylbenzene containing phenylmethylamine groups, used as solid support for peptide synthesis, was examined regarding physicochemical and anion exchanger chromatographic properties. The greater the ionic strength of the medium the poorer the solvation of beads. This

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service