Skip to Content
Merck
All Photos(1)

Key Documents

536725

Sigma-Aldrich

PAMAM dendrimer, ethylenediamine core, generation 7.0 solution

5 wt. % in methanol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[NH2(CH2)2NH2]:(G=7);dendri PAMAM(NH2)512
CAS Number:
Molecular Weight:
116488.71
MDL number:
UNSPSC Code:
12162002
NACRES:
NA.23
Pricing and availability is not currently available.

description

amino surface groups

form

liquid

mol wt

generation 7.0

feature

core type ethylenediamine core (2-carbon core)

concentration

5 wt. % in methanol

no. Surface Groups

512

refractive index

n20/D 1.341

density

0.8 g/mL at 25 °C

storage temp.

2-8°C

Application


  • Development of Crystalline Cu(2)S Nanowires via a Direct Synthesis Process and Its Potential Applications.: This research explores the synthesis of crystalline Cu(2)S nanowires, highlighting their potential applications in nanotechnology, possibly including energy storage, solar cells, and photocatalytic systems. The study focuses on direct synthesis methods that could pave the way for new uses in high-efficiency energy solutions and other advanced material applications (Chen et al., 2020).


Physical form

Contains 512 surface primary amino groups

Legal Information

Manufactured by Dendritech® , Inc.
Dendritech is a registered trademark of Dendritech, Inc.

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Flam. Liq. 2 - STOT SE 1

Target Organs

Eyes,Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

49.5 °F

Flash Point(C)

9.7 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yubo Qin et al.
Colloids and surfaces. B, Biointerfaces, 127, 247-255 (2015-02-18)
A new strategy to prepare an ultra non-fouling SU-8/poly(dimethylsiloxane) (PDMS) hybrid microfluidic system is presented. We report a simple method to bond both SU-8 and PDMS surfaces to prepare the hybrid microfluidic device and then simultaneously modify both SU-8 and

Articles

Replace volatile organic solvents with water due to environmental concerns, overcoming challenges in solubility.

Replace volatile organic solvents with water due to environmental concerns, overcoming challenges in solubility.

Replace volatile organic solvents with water due to environmental concerns, overcoming challenges in solubility.

Replace volatile organic solvents with water due to environmental concerns, overcoming challenges in solubility.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service