53530
DL-Homocysteine thiolactone hydrochloride
≥99.0% (AT)
Synonym(s):
DL-2-Amino-4-mercaptobutyric acid 1,4-thiolactone hydrochloride
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C4H7NOS · HCl
CAS Number:
Molecular Weight:
153.63
Beilstein:
3689338
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
42021308
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Quality Level
Assay
≥99.0% (AT)
form
solid
loss
≤0.5% loss on drying
mp
202 °C (dec.) (lit.)
~203 °C (dec.)
functional group
thioester
SMILES string
Cl[H].NC1CCSC1=O
InChI
1S/C4H7NOS.ClH/c5-3-1-2-7-4(3)6;/h3H,1-2,5H2;1H
InChI key
ZSEGSUBKDDEALH-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
DL-Homocysteine thiolactone hydrochloride (HTL-HCl) is a cyclic amino acid derivative that exhibits root-growth inhibitory activity.
Application
DL-Homocysteine thiolactone hydrochloride may be used in the preparation of:
- DL-buthionine
- DL-homocysteine
- poly[α,β-(N-2-aminoethyl-dl-aspartamide)]-poly[α,β-(N-2-hydroxyethyl-dl-aspartamide)]-poly[α,β-(N-3-mercapto-1-methoxycarbonyl-propyl-dl-aspartamide)] copolymer diclofenac conjugate (PAHMA-Dic)
- bis[ S-(dimethylarsino)homocysteine]
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Joanna Malinowska et al.
Platelets, 22(4), 277-283 (2011-05-12)
Resveratrol (3,4',5-trihydroxystilben), a phenolic antioxidant synthesized in grapes and other plants, and also present in wine, has been suggested to help prevent cardiovascular events. In this study the influence of resveratrol on platelet aggregation during a model of hyperhomocysteinemia was
Jerott R Moore et al.
Journal of neuroimmunology, 324, 100-114 (2018-09-30)
We investigated how one calcitriol dose plus vitamin D3 reverses experimental autoimmune encephalomyelitis (EAE), a multiple sclerosis model. This protocol rapidly increased CD4+ T cell Ikzf2 transcripts, Helios protein, and CD4+Helios+FoxP3+ T regulatory cells. It also rapidly increased CD4+ T
Reza Yousefi et al.
International journal of biological macromolecules, 51(3), 291-298 (2012-05-29)
In the current study both structural alteration and fibrillation of insulin were studied in the presence of homocysteine thiolactone (HCTL). The spectroscopic studies revealed that HCTL increases rate of insulin unfolding, giving rise to the appearance of solvent-exposed hydrophobic regions
Hieronim Jakubowski et al.
Advances in clinical chemistry, 55, 81-103 (2011-12-01)
Protein-related homocysteine (Hcy) metabolism produces Hcy-thiolactone, N-Hcy-protein, and N epsilon-homocysteinyl-lysine (N epsilon-Hcy-Lys). Hcy-thiolactone is generated in an error-editing reaction in protein biosynthesis when Hcy is erroneously selected in place of methionine by methionyl-tRNA synthetase. Hcy-thiolactone, an intramolecular thioester, is chemically
O W Griffith et al.
The Journal of biological chemistry, 254(16), 7558-7560 (1979-08-25)
Buthionine sulfoximine (S-n-butyl homocysteine sulfoximine), the most potent of a series of analogs of methionine sulfoximine thus far studied (Griffith, O.W., Anderson, M.E., and Meister, A. (1979) J. Biol. Chem. 254, 1205-1210), inhibited gamma-glutamylcysteine synthetase about 20 times more effectively
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service