Skip to Content
MilliporeSigma

Skip To

44925

DL-Homocysteine

analytical standard

Synonym(s):

2-Amino-4-mercaptobutyric acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice
25 mg

Available to ship TODAYfromMILWAUKEE

$91.20

About This Item

Linear Formula:
HSCH2CH2CH(NH2)COOH
CAS Number:
Molecular Weight:
135.18
UNSPSC Code:
85151701
NACRES:
NA.24
PubChem Substance ID:
EC Number:
207-222-9
Beilstein/REAXYS Number:
1721683
MDL number:

$91.20


Available to ship TODAYDetails


Request a Custom Order
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

analytical standard

Quality Segment

assay

≥95% (T)

optical activity

[α]/D 0±1°, c = 1 in H2O

shelf life

limited shelf life, expiry date on the label

analyte chemical class(es)

amino acids, peptides, proteins

technique(s)

HPLC: suitable

color

white to off-white

mp

232-233 °C (lit.)

application(s)

food and beverages

format

neat

storage temp.

−20°C

SMILES string

NC(CCS)C(O)=O

InChI

1S/C4H9NO2S/c5-3(1-2-8)4(6)7/h3,8H,1-2,5H2,(H,6,7)

InChI key

FFFHZYDWPBMWHY-UHFFFAOYSA-N

Application

Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.
DL-Homocysteine may be used as an analytical reference standard for the quantification of homocysteine in biological samples using different chromatography techniques.

Biochem/physiol Actions

Increased plasma homocysteine is a risk factor for coronary heart disease and carcinogenesis; may be a marker for tumor cell growth.

Analysis Note

identification (LS-MS): identity conforms

Disclaimer

store under argon

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
73767PHR1241H4628
format

neat

format

neat

format

neat

format

-

technique(s)

HPLC: suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

HPLC: suitable, gas chromatography (GC): suitable

technique(s)

-

grade

analytical standard

grade

certified reference material, TraceCERT®

grade

certified reference material, pharmaceutical secondary standard

grade

-

shelf life

limited shelf life, expiry date on the label

shelf life

-

shelf life

-

shelf life

-

assay

≥95% (T)

assay

-

assay

-

assay

≥95% (titration)

storage temp.

−20°C

storage temp.

-

storage temp.

2-30°C

storage temp.

−20°C


Still not finding the right product?

Explore all of our products under DL-Homocysteine


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters