Recommended Products
Quality Level
Assay
97%
mp
116-119 °C (lit.)
functional group
carboxylic acid
iodo
SMILES string
OC(=O)Cc1ccccc1I
InChI
1S/C8H7IO2/c9-7-4-2-1-3-6(7)5-8(10)11/h1-4H,5H2,(H,10,11)
InChI key
IUHXGZHKSYYDIL-UHFFFAOYSA-N
Related Categories
General description
2-Iodophenylacetic acid is a substituted acetic acid that can be prepared from 2-iodobenzyl cyanide. It undergoes palladium-catalyzed reaction with allenes to form 1,3-butadienes. 2-Iodophenylacetic acid also undergoes photolysis in carbon tetrachloride to form the corresponding chloro compound.
Application
2-Iodophenylacetic acid may be used in the preparation of:
- 2-iodophenylacetyl chloride
- 3,4-dimethoxyphenyl 2-iodophenylacetate
- N-{2-[2-(2-iodophenyl)-acetylamino]-ethyl}-2-(3,4- dichlorophenyl)-acetamide
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Regioselective Synthesis of Isoquino [1,2-b][3] benzazepines (Homoprotoberberines) through 11-Membered-Ring Stilbene Lactams Obtained by Radical Macrocyclization.
The Journal of Organic Chemistry, 64(13), 4830-4833 (1999)
Photochemical synthesis of aromatic chloro compounds from aromatic iodo compounds.
The Journal of Organic Chemistry, 35(2), 528-529 (1970)
Synthesis of benzo[c]chromen-6-ones via novel cyclic aryl?Pd (II)?ester enolate intermediates.
Tetrahedron, 63(45), 10889-10895 (2007)
Pd?Catalyzed Reactions of Allenylphosphonates and Related Allenes with Functionalized 2?Iodophenols, 2?Iodobenzoic Acid, and 2?Iodobenzyl Alcohol Leading to Functionalized Benzofurans, Isocoumarins, and Benzopyrans.
European Journal of Organic Chemistry, 5927-5940 (2009)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service