Skip to Content
Merck
All Photos(1)

Key Documents

529257

Sigma-Aldrich

Poly(2-hydroxyethyl methacrylate)

average Mv 1,000,000

Synonym(s):

Poly(2-HEMA), Poly-HEMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O3)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

crystals

Quality Level

mol wt

average Mv 1,000,000

density

1.15 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OCCO

InChI

1S/C6H10O3/c1-5(2)6(8)9-4-3-7/h7H,1,3-4H2,2H3

InChI key

WOBHKFSMXKNTIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Physical form

Water-swellable polymer. Hydrogel.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

N Nogueira et al.
Journal of colloid and interface science, 385(1), 202-210 (2012-07-31)
The behavior of poly(2-hydroxyethyl methacrylate) (PHEMA) polymer monolayer spread on water was studied under various experimental conditions. The influence of subphase pH and temperature, compression speed, elapsed time from the deposit of the monolayer and the recording of the surface
Amit D Bhrany et al.
JAMA facial plastic surgery, 15(1), 29-33 (2013-01-19)
To evaluate the performance of a sphere-templated poly(2-hydroxyethyl methacrylate) (poly[HEMA]) tissue scaffold as a subcutaneous implant by comparing it with widely used high-density porous polyethylene (HDPPE) implant material. We implanted sphere-templated porous poly-(HEMA) and HDPPE disks into the dorsal subcutis
Olga Samsonova et al.
Acta biomaterialia, 9(2), 4994-5002 (2012-10-13)
The mechanism causing variability in DNA transfection efficacy for low-molecular-weight pDMAEMA (poly(2-(dimethylamino)ethyl methacrylate) and pDMAEMA-b-pHEMA (poly(2-(dimethyl amino)ethylmethacrylate)-block-poly(2-hydroxyl methacrylate)) has so far remained unclear, apart from the evidence of beneficial effects of the pHEMA grafting. This study has explicitly characterized the
Yunfeng Li et al.
ACS applied materials & interfaces, 5(6), 2126-2132 (2013-02-23)
This paper presents a versatile way to prepare multiscale and gradient patterns of proteins. The protein patterns are fabricated by conjugating proteins covalently on patterns of polymer brush that are prepared by techniques combining colloidal lithography with photolithography, and two-step
Craig N Czyz et al.
Ophthalmic plastic and reconstructive surgery, 29(1), e25-e27 (2012-07-28)
Hydrogel was a commonly used material for scleral buckling in the early 1980s to the mid-1990s. Use of hydrogel ceased due to a high complication rate, including frequent migration. Various symptoms and clinical findings have been reported with hydrogel migration.

Articles

Professor Shrike Zhang (Harvard Medical School, USA) discusses advances in 3D-bioprinted tissue models for in vitro drug testing, reviews bioink selections, and provides application examples of 3D bioprinting in tissue model biofabrication.

Professor Shrike Zhang (Harvard Medical School, USA) discusses advances in 3D-bioprinted tissue models for in vitro drug testing, reviews bioink selections, and provides application examples of 3D bioprinting in tissue model biofabrication.

Professor Shrike Zhang (Harvard Medical School, USA) discusses advances in 3D-bioprinted tissue models for in vitro drug testing, reviews bioink selections, and provides application examples of 3D bioprinting in tissue model biofabrication.

Professor Shrike Zhang (Harvard Medical School, USA) discusses advances in 3D-bioprinted tissue models for in vitro drug testing, reviews bioink selections, and provides application examples of 3D bioprinting in tissue model biofabrication.

See All

Protocols

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service