Skip to Content
Merck
All Photos(1)

Key Documents

D7290

Sigma-Aldrich

Deoxyribonucleic acid from herring sperm

single-stranded for hybridization

Synonym(s):

DNA

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
41106310
eCl@ss:
32160414
NACRES:
NA.52

biological source

(herring sperm)

Quality Level

grade

for molecular biology

form

solution

shipped in

dry ice

storage temp.

−20°C

General description

Suitable for use as a blocking agent in Southern hybridizations.

Application

Deoxyribonucleic acid from herring sperm is suitable for use as a blocking agent in Southern hybridizations. It was used in preparation of brain sections of mice for BrdU immunohistochemistry. It was also used for hybridization reaction of dot blot membranes.
In order to decrease any non-specific hybridization of the probe to a substrate, blocking agents must be used. Generally, a combination of blocking reagent, detergent, and denatured, fragmented DNA is used to accomplish this. Sigma offers sonicated, denatured DNA from a variety of species for use as a blocking agent in Northern and Southern blotting and other nucleic acid hybridization techniques.
Many factors contribute to the signal-to-noise ratio in nucleic acid hybridizations. These factors include the presence of solvent (formamide), hybridization temperature, length of hybridization, volume of hybridization solution, degree and method of agitation, use of blocking reagents, concentration and specific activity of the probe, use of molecular agents to increase the rate of nucleic acid reassociation, and the degree of stringency used during the washing of the membrane.

In order to decrease any non-specific hybridization of the probe to a substrate, blocking agents must be used. Generally, a combination of blocking reagent, detergent, and denatured, fragmented DNA is used to accomplish this. Sigma offers sonicated, denatured DNA from a variety of species for use as a blocking agent in Northern and Southern blotting and other nucleic acid hybridization techniques.

Preparation Note

This DNA is phenol-chloroform extracted, ethanol precipitated, and sonicated to produce single-stranded fragments which comigrate with the 587 and 831 base pair marker fragments.

Other Notes

DNA in solution will reanneal on standing at room temperature so it is recommended to boil the solution for 10 minutes and then cool on ice for at least 5 minutes prior to use.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marloes Prins et al.
PloS one, 8(12), e83835-e83835 (2014-01-01)
Multiple sclerosis (MS) has been mainly attributed to white matter (WM) pathology. However, recent evidence indicated the presence of grey matter (GM) lesions. One of the principal mediators of inflammatory processes is interleukin-1β (IL-1β), which is known to play a
Hanbin Ma et al.
Scientific reports, 3, 2730-2730 (2013-09-26)
Herein, we describe a novel integrated biosensor for performing dielectric spectroscopy to analyze biological samples. We analyzed biomolecule samples with different concentrations and demonstrated that the solution's impedance is highly correlated with the concentration, indicating that it may be possible
Simon A Joosse et al.
BMC cancer, 7, 43-43 (2007-03-09)
Array Comparative Genomic Hybridization (aCGH) is a rapidly evolving technology that still lacks complete standardization. Yet, it is of great importance to obtain robust and reproducible data to enable meaningful multiple hybridization comparisons. Special difficulties arise when aCGH is performed
Minna M Poranen et al.
Journal of virology, 80(16), 8081-8088 (2006-07-29)
Virus-induced changes in cellular gene expression and host physiology have been studied extensively. Still, there are only a few analyses covering the entire viral replication cycle and whole-host gene pool expression at the resolution of a single gene. Here we
Philip Thomas et al.
Mutagenesis, 23(1), 57-65 (2007-12-01)
Alzheimer's disease (AD) is a premature ageing syndrome characterized by cognitive impairment arising from neuropathological changes occurring within specific areas of the brain. We report a 1.5-fold increase in trisomy 21 (P < 0.001) and a 1.2-fold increase in trisomy

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service