Skip to Content
Merck
All Photos(3)

Key Documents

525057

Sigma-Aldrich

4-Pyrazoleboronic acid pinacol ester

97%

Synonym(s):

4,4,5,5-Tetramethyl-2-(1H-pyrazol-2-yl)-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-(1H-pyrazol-4-yl)-1,3,2-dioxaborolane, 4,4,5,5-Tetramethyl-2-(pyrazol-4-yl)-1,3,2-dioxaborolane, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole, Pyrazol-4-ylboronic acid pinacol ester

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)4C2O2BC3N2H3
CAS Number:
Molecular Weight:
194.04
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

142-146 °C (lit.)

SMILES string

CC1(C)OB(OC1(C)C)c2cn[nH]c2

InChI

1S/C9H15BN2O2/c1-8(2)9(3,4)14-10(13-8)7-5-11-12-6-7/h5-6H,1-4H3,(H,11,12)

InChI key

TVOJIBGZFYMWDT-UHFFFAOYSA-N

Application

Reagent used for
  • Suzuki-Miyaura cross-couplings
  • Ruthenium-catalyzed asymmetric hydrogenation

Reagent used in preparation of inhibitors of many highly significant therapeutic enzymes and kinases containing the privileged scaffold pyrazole, including
  • VEGF
  • Aurora
  • Rho (ROCK)
  • Janus Kinase 2 (JAK)
  • c-MET
  • ALK
  • S-nitrosoglutathione reductase
  • CDC7
  • Acetyl-CoA carboxylase
  • Prosurvival Bcl-2 protein
  • Viral RNA-Dependent RNA polymerase
  • Long Chain Fatty Acid Elongase 6
  • PI3
  • AKT
  • Chk1
  • Protein Kinase B

Legal Information

Product of Boron Molecular

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Robert M Garbaccio et al.
Bioorganic & medicinal chemistry letters, 17(22), 6280-6285 (2007-09-29)
From HTS lead 1, a novel benzoisoquinolinone class of ATP-competitive Chk1 inhibitors was devised and synthesized via a photochemical route. Using X-ray crystallography as a guide, potency was rapidly enhanced through the installation of a tethered basic amine designed to
Dirk A Heerding et al.
Journal of medicinal chemistry, 51(18), 5663-5679 (2008-09-20)
Overexpression of AKT has an antiapoptotic effect in many cell types, and expression of dominant negative AKT blocks the ability of a variety of growth factors to promote survival. Therefore, inhibitors of AKT kinase activity might be useful as monotherapy
Design and evaluation of 3,6-di(hetero)aryl imidazo[1,2-a]pyrazines as inhibitors of checkpoint and other kinases
Matthews, T. P.; et al.
Bioorganic & Medicinal Chemistry, 20, 4045-4049 (2010)
Christoffer Bengtsson et al.
Bioorganic & medicinal chemistry, 19(10), 3039-3053 (2011-04-26)
Inhibition of acetyl-CoA carboxylases has the potential for modulating long chain fatty acid biosynthesis and mitochondrial fatty acid oxidation. Hybridization of weak inhibitors of ACC2 provided a novel, moderately potent but lipophilic series. Optimization led to compounds 33 and 37
Benjamin Perry et al.
Bioorganic & medicinal chemistry letters, 18(19), 5299-5302 (2008-09-09)
Optimization of the cellular and pharmacological activity of a novel series of PI3 kinase inhibitors targeting multiple isoforms is described.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service