Skip to Content
Merck
All Photos(1)

Key Documents

518786

Sigma-Aldrich

4-Hydroxy-3-methoxyphenylboronic acid pinacol ester

98%

Synonym(s):

2-Methoxy-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenol, 4-Hydroxy-3-methoxyphenylboronic acid, pinacol cyclic ester

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H19BO4
CAS Number:
Molecular Weight:
250.10
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

mp

105-109 °C (lit.)

SMILES string

COc1cc(ccc1O)B2OC(C)(C)C(C)(C)O2

InChI

1S/C13H19BO4/c1-12(2)13(3,4)18-14(17-12)9-6-7-10(15)11(8-9)16-5/h6-8,15H,1-5H3

InChI key

WFSJROCEOJANPD-UHFFFAOYSA-N

Application

4-Hydroxy-3-methoxyphenylboronic acid pinacol ester can be used as a reactant:
  • In the Suzuki-Miyaura cross-coupling reaction.
  • To prepare ethylidenedihydroindolones as potential neoplastic agents.
  • To synthesize 5,6-dihydropyrrolo[2,1-b]isoquinoline derivatives, which can be utilized as scaffolds to prepare lamellarin analogs via regioselective bromination and Suzuki cross-coupling reactions.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

5, 6-Dihydropyrrolo [2, 1-b] isoquinolines as scaffolds for synthesis of lamellarin analogues
Olsen CA, et al.
Tetrahedron Letters, 46(12), 2041-2044 (2005)
New oxidative labels for electrochemical detection of DNA
Simonova A, et al.
Collection of Czechoslovak Chemical Communications, 14, 365-366 (2015)
Synthesis and biological evaluation of 3-ethylidene-1, 3-dihydro-indol-2-ones as novel checkpoint 1 inhibitors
L Nan-Horng, et al.
Bioorganic & Medicinal Chemistry Letters, 16(2), 421-426 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service