Skip to Content
Merck
All Photos(2)

Key Documents

515027

Sigma-Aldrich

4-Methyl-1-piperazinecarbonyl chloride hydrochloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11ClN2O · HCl
CAS Number:
Molecular Weight:
199.08
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

225-228 °C (lit.)

SMILES string

Cl[H].CN1CCN(CC1)C(Cl)=O

InChI

1S/C6H11ClN2O/c1-8-2-4-9(5-3-8)6(7)10/h2-5H2,1H3

InChI key

FBAIGEMWTOSCRU-UHFFFAOYSA-N

Application

4-Methyl-1-piperazinecarbonyl chloride hydrochloride may be used in the synthesis of tert-butyl 2-(N-ethyl-4-methylpiperzaine-1-carboxamido)ethylcarbamate and 4b,8,8-trimethyl-9,10-dioxo-4b,5,6,7,8,8a,9,10-octahydrophenanthren-2-yl 4-methyl-piperazine-1-carboxylate.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amy Junnila et al.
Bioconjugate chemistry, 18(6), 1818-1823 (2007-10-12)
New fluorescein and rhodamine B-labeled antifilarial drug DEC analogues for use in drug localization studies with confocal microscopy have been prepared by a high-yield three-step synthesis. The resulting beta-amine-substituted DEC analogue has a single ethyl substituent which is beta-aminated to
Qiong Wei et al.
PloS one, 7(12), e52199-e52199 (2013-01-10)
Thymidine kinases (TKs) have been considered one of the potential targets for anticancer therapeutic because of their elevated expressions in cancer cells. However, nucleobase analogs targeting TKs have shown poor selective cytotoxicity in cancer cells despite effective antiviral activity. 3'-Deoxythymidine
Brown DJ.
The Chemistry of Heterocyclic Compounds, 317-317 (2003)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service