Skip to Content
Merck
All Photos(2)

Key Documents

365386

Sigma-Aldrich

N,N-Diisopropylcarbamoyl chloride

98%

Synonym(s):

Diisopropylcarbamic chloride, Diisopropylcarbamyl chloride, N,N-Bis(1-methylethyl)carbamic chloride

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2CH]2NCOCl
CAS Number:
Molecular Weight:
163.65
Beilstein:
1754834
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

bp

90-93 °C/15 mmHg (lit.)

mp

57-59 °C (lit.)

SMILES string

CC(C)N(C(C)C)C(Cl)=O

InChI

1S/C7H14ClNO/c1-5(2)9(6(3)4)7(8)10/h5-6H,1-4H3

InChI key

RSAFAYLZKCYUQW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N,N-Diisopropylcarbamoyl chloride may be used in the preparation of:
  • 1-aryl-1-alkenyl N,N-diisopropylcarbamates
  • 1,3-diphenylallyl carbamate
  • (Z)-1,3-diphenyl-1-propenyl N,N-diisopropylcarbamate
  • (R)-1-(2-methoxy-4-methylphenyl)ethyl diisopropyl-carbamate

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Asymmetric ?-Deprotonation and Substitution Reactions of (Z)-1, 3-Diphenyl-1-propenyl N, N-Diisopropylcarbamate.
Reuber J, et al.
European Journal of Organic Chemistry, 14, 3017-3025 (2005)
The total synthesis of (-)-aplysin via a lithiation-borylation-propenylation sequence.
Fletcher CJ, et al.
Tetrahedron, 68(23), 7598-7604 (2012)
Stereoselective Intermolecular Carbolithiation of Open-Chain and Cyclic 1-Aryl-1-alkenyl N,N-Diisopropylcarbamates Coupled with Electrophilic Substitution. Observation of p-Carboxylation in a Benzyllithium Derivative.
Peters JG, et al.
Synthesis, 03, 0381-0292 (2002)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service