511692
Sodium 2-ethylhexanoate
97%
Synonym(s):
2-Ethylcaproic acid sodium salt, Sodium-2-ethylcaproate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
CH3(CH2)3CH(C2H5)CO2Na
CAS Number:
Molecular Weight:
166.19
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23
Recommended Products
Quality Level
Assay
97%
form
powder, crystals or chunks
reaction suitability
core: sodium
mp
>300 °C (lit.)
SMILES string
[Na+].CCCCC(CC)C([O-])=O
InChI
1S/C8H16O2.Na/c1-3-5-6-7(4-2)8(9)10;/h7H,3-6H2,1-2H3,(H,9,10);/q;+1/p-1
InChI key
VYPDUQYOLCLEGS-UHFFFAOYSA-M
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Repr. 1B
Storage Class Code
6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Meeri Visnapuu et al.
Colloids and surfaces. B, Biointerfaces, 169, 222-232 (2018-05-21)
Application of efficient antimicrobial surfaces has been estimated to decrease both, the healthcare-associated infections and the spread of antibiotic-resistant bacteria. In this paper, we prepared ZnO and ZnO/Ag nanoparticle covered surfaces and evaluated their antimicrobial efficacy towards a Gram-negative bacterial
Fabrizio Carta et al.
Bioorganic & medicinal chemistry letters, 21(8), 2521-2526 (2011-03-16)
The inhibition of the β-carbonic anhydrases (CAs, EC 4.2.1.1) from the pathogenic fungi Cryptococcus neoformans (Can2) and Candida albicans (Nce103) with a series of 25 branched aliphatic and aromatic carboxylates has been investigated. Human isoforms hCA I and II were
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service