Skip to Content
Merck
All Photos(1)

Key Documents

51053

Sigma-Aldrich

1-Ethyl-3-methylimidazolium acetate

≥95.0% (HPLC)

Synonym(s):

EMIM Ac

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H14N2O2
CAS Number:
Molecular Weight:
170.21
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥95.0% (HPLC)

origin

Switzerland origin

mp

>30 °C (product can occur as an undercooled melt)

density

1.101 g/cm3 at 20 °C

SMILES string

CC([O-])=O.CCn1cc[n+](C)c1

InChI

1S/C6H11N2.C2H4O2/c1-3-8-5-4-7(2)6-8;1-2(3)4/h4-6H,3H2,1-2H3;1H3,(H,3,4)/q+1;/p-1

InChI key

XIYUIMLQTKODPS-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

General description

1-Ethyl-3-methylimidazolium acetate ([EMIM][Ac]) is a water miscible, ecofriendly imidazolium ionic liquid. It shows good cellulose dissolution ability. Study on the influence of [EMIM]Ac on the growth of Clostridium sp. and Pseudomonas putida showed hormetic effect. Addition of inorganic salts has been reported to enhance its ionicity. The thermal stability of cellulose in [EMIM]Ac has been investigated.

Application

1-Ethyl-3-methylimidazolium acetate is suitable for use as cellulose solvent in the homogeneous conversion of cellulose with acid chlorides, trityl chloride and tosyl chloride. It may be employed as solvent for the synthesis of copolymer of starch grafted with polystyrene (starch-g-PS) and environmentally friendly cellulose films.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2 - Skin Sens. 1B

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

327.2 °F - closed cup

Flash Point(C)

164 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Koel Saha et al.
3 Biotech, 8(8), 374-374 (2018-08-15)
The primary focus of this work was to recover lignin and investigate the structural changes in sugarcane bagasse after pretreatment with ionic liquid 1-ethyl-3-methylimidazolium acetate ([EMIM]oAc). 90% lignin recovery was achieved while bagasse was treated with [EMIM]oAc at 140 °C, 120 min
Anne Michud et al.
Carbohydrate polymers, 117, 355-363 (2014-12-17)
The thermal stability of cellulose in the ionic liquid (IL) 1-ethyl-3-methylimidazolium acetate, [emim]OAc was investigated. For this purpose, Eucalyptus urugrandis prehydrolysis kraft pulp was first dissolved in [emim]OAc by means of a vertical kneader and then stored at three different
Interactions of Ionic Liquids with Polysaccharides 1. Unexpected Acetylation of Cellulose with 1-Ethyl-3-methylimidazolium Acetate.
Kohler S, et al.
Macromolecular Rapid Communications, 28(24), 2311-2317 (2007)
Boxuan Li et al.
Bioactive materials, 5(3), 577-583 (2020-05-15)
Chitosan is a nature-based polymer with low toxicity, excellent biocompatibility and biodegradability. However, the intractable solubility of chitosan in water and most conventional solvents hampers its biomedical applications. Following the dissolution method for dissolving chitosan in plain water developed by
Yongjun Men et al.
Carbohydrate polymers, 121, 348-354 (2015-02-11)
The copolymer of starch grafted with polystyrene (starch-g-PS) was synthesized with high grafting percentage by utilizing the ionic liquid 1-ethyl-3-methylimidazolium acetate ([EMIM]Ac) as solvent and potassium persulfate as initiator. The effect of various parameters upon the polymerization were studied including:

Articles

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

BASIONICS™ Ionic Liquids offer diverse properties for varied applications in the BASF portfolio.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service