298212
Copper(I) thiocyanate
99%
Synonym(s):
Cuprous thiocyanate
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Recommended Products
Assay
99%
form
powder
application(s)
battery manufacturing
SMILES string
[Cu]SC#N
InChI
1S/CHNS.Cu/c2-1-3;/h3H;/q;+1/p-1
InChI key
PDZKZMQQDCHTNF-UHFFFAOYSA-M
Related Categories
General description
The crystal belongs to the orthorhombic system, space group: Pbca,a=10.994(9)Å, b=7.224(3)Å,c=6.662(2)Å.
Application
The product can be prepared from Cu(CH3COO)2·H2O, AgNO3, NH4NCS and im(im = imidazole). The structure and optical properties were reported. It is an air- and light-stable source of Cu(I).
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Acute 1 - Aquatic Chronic 1
Supplementary Hazards
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of human nutrition, 34(6), 445-449 (1980-12-01)
The advantages and disadvantages of faecal markers that are available in the UK are discussed. Opinions and comments from units using them are included.
Synthesis, structure and non-linear optical property of a copper(II) thiocyanate three-dimensional supramolecular compound
Journal of Molecular Structure, 660, 107-112 (2003)
Dalton transactions (Cambridge, England : 2003), (39)(39), 5290-5292 (2008-10-02)
The role of short-range diffusion in solvent-assisted mechanochemical synthesis is demonstrated in studies of a polymorphic transition and a ligand dissociation reaction involving copper(I) thiocyanate complexes.
The New Zealand medical journal, 93(676), 36-38 (1981-01-28)
Faecal fat excretion was estimated in 24 patients using three methods. Quantitative estimations of fat excretion were calculated from both a three day faecal fat collection and a twenty-four hour faecal collection corrected for excretion of a cuprous thiocyanate marker.
Dalton transactions (Cambridge, England : 2003), 41(25), 7513-7525 (2012-05-24)
A number of adducts of copper(I) thiocyanate with bulky tertiary phosphine ligands, and some nitrogen-base solvates, were synthesized and structurally and spectroscopically characterised. CuSCN:PCy3 (1:2), as crystallized from pyridine, is shown by a single crystal X-ray study to be a
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service