Skip to Content
Merck
All Photos(2)

Key Documents

482277

Sigma-Aldrich

Lithium manganese(III,IV) oxide

greener alternative

electrochemical grade

Synonym(s):

LMO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
LiMn2O4
CAS Number:
Molecular Weight:
180.81
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
grade:
electrochemical grade
form:
powder

grade

electrochemical grade

Quality Level

form

powder

greener alternative product characteristics

Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

concentration

59.8-61.7% Mn (by KMnO4, titration)

impurities

≤20000 ppm Trace Metal Analysis

particle size

<5 μm

mp

>400 °C (lit.)

application(s)

battery manufacturing

greener alternative category

SMILES string

[Li+].[O-][Mn]=O.O=[Mn]=O

InChI

1S/Li.2Mn.4O/q+1;;;;;;-1

InChI key

VLXXBCXTUVRROQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Lithium manganese(III,IV) oxide (LMO) is a class of electrode material that can be used in the fabrication of lithium-ion batteries. Lithium-ion batteries consist of anode, cathode, and electrolyte with a charge-discharge cycle. These materials enable the formation of greener and sustainable batteries for electrical energy storage.
We are committed to bringing you Greener Alternative Products, which adhere to one or more of The 12 Principles of Greener Chemistry. This product has been enhanced for energy efficiency. Find details here.

Application

Lithium manganese(III,IV) oxide, a low cost and eco-friendly cubic spinel structure, can be used as a cathode material for the formation of high-performance lithium-ion batteries. It can also be used as a mild oxidizing agent for the production of dopaminequinone.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A LiMn2O4 based electrochemical scheme for selective measurement of dopamine
Leu HJ and Lin MS
Electroanalysis, 18(3), 307-310 (2006)
Spinel LiMn2O4 nanorods as lithium ion battery cathodes
Kim DK, et al.
Nano Letters, 8(11), 3948-3952 (2008)
Oxygen-Vacancy Ordering at Surfaces of Lithium Manganese (III, IV) Oxide Spinel Nanoparticles
Huang R, et al.
Angewandte Chemie (International ed. in English), 50(13), 3053-3057 (2011)
Nicole L Green et al.
Soft matter, 11(27), 5523-5530 (2015-06-13)
Oil migration in chocolate and chocolate-based confections leads to undesirable visual and textural changes. Establishing ways to slow this unavoidable process would increase shelf life and reduce consumer rejection. Diffusion is most often credited as the main pathway by which
Brenda Hanna-Pladdy et al.
Journal of the International Neuropsychological Society : JINS, 21(4), 259-270 (2015-04-30)
Parkinson's disease (PD) is characterized by asymmetric motor symptom onset attributed to greater degeneration of dopamine neurons contralateral to the affected side. However, whether motor asymmetries predict cognitive profiles in PD, and to what extent dopamine influences cognition remains controversial.

Articles

Nanomaterials for Energy Storage in Lithium-ion Battery Applications

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

HEVs address rising fuel costs and emissions concerns, utilizing battery packs alongside internal combustion engines for enhanced performance.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service