Skip to Content
Merck
All Photos(1)

Key Documents

480479

Sigma-Aldrich

Bromocyclohexane-d11

≥98 atom % D, ≥98% (CP)

Synonym(s):

Cyclohexyl bromide-d11

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6D11Br
CAS Number:
Molecular Weight:
174.12
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:
NACRES:
NA.12
Pricing and availability is not currently available.

isotopic purity

≥98 atom % D

Quality Level

Assay

≥98% (CP)

refractive index

n20/D 1.492 (lit.)

bp

166-167 °C (lit.)

density

1.412 g/mL at 25 °C (lit.)

application(s)

pharmaceutical

mass shift

M+11

SMILES string

[2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])(Br)C([2H])([2H])C1([2H])[2H]

InChI

1S/C6H11Br/c7-6-4-2-1-3-5-6/h6H,1-5H2/i1D2,2D2,3D2,4D2,5D2,6D

InChI key

AQNQQHJNRPDOQV-KAFHOZLVSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service