Skip to Content
Merck
All Photos(1)

Documents

B56501

Sigma-Aldrich

4-Bromoanisole

≥99.0%

Synonym(s):

1-Bromo-4-methoxybenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4OCH3
CAS Number:
Molecular Weight:
187.03
Beilstein:
1237590
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99.0%

form

liquid

refractive index

n20/D 1.564 (lit.)

bp

223 °C (lit.)

mp

9-10 °C (lit.)

density

1.494 g/mL at 25 °C (lit.)

SMILES string

COc1ccc(Br)cc1

InChI

1S/C7H7BrO/c1-9-7-4-2-6(8)3-5-7/h2-5H,1H3

InChI key

QJPJQTDYNZXKQF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Bromoanisole is a useful brominating reagent. It is formed as reaction product in the reaction between HOBr and anisole. Suzuki coupling of 4-bromoanisole with phenylboronic acid catalyzed by palladium pincer complexes has been studied. Heck Reaction of 4-bromoanisole with ethyl acrylates in room-temperature ionic liquids is reported to afford ethyl 4-methoxycinnamate.

Application

4-Bromoanisole was used in the synthesis of aryl 1,3-diketones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

201.2 °F

Flash Point(C)

94 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Amy E Bryant et al.
PloS one, 12(2), e0172486-e0172486 (2017-03-01)
Acute muscle injuries are exceedingly common and non-steroidal anti-inflammatory drugs (NSAIDs) are widely consumed to reduce the associated inflammation, swelling and pain that peak 1-2 days post-injury. While prophylactic use or early administration of NSAIDs has been shown to delay
Yasuyuki Nakamura et al.
Applied and environmental microbiology, 84(15) (2018-05-29)
The methylotrophic yeast Pichia pastoris is widely used to produce recombinant proteins, taking advantage of this species' high-density cell growth and strong ability to secrete proteins. Circular plasmids containing the P. pastoris-specific autonomously replicating sequence (PARS1) permit transformation of P.
Charles E Wood et al.
Carcinogenesis, 36(7), 782-791 (2015-04-29)
Environmental exposures occurring early in life may have an important influence on cancer risk later in life. Here, we investigated carryover effects of dichloroacetic acid (DCA), a small molecule analog of pyruvate with metabolic programming properties, on age-related incidence of
J Vokurka et al.
Polish journal of veterinary sciences, 23(2), 169-176 (2020-07-07)
Different approaches to enhance healing of hard or soft tissues include the use of cytokines and growth factors to modify cellular behaviour. Numerous growth factors are found in autologous blood concentrates - platelet-rich plasma (PRP) and platelet-rich fibrin (PRF). Enamel
John D Sivey et al.
Environmental science & technology, 49(8), 4937-4945 (2015-03-25)
When bromide-containing waters are chlorinated, conventional wisdom typically assumes HOBr is the only active brominating agent. Several additional and often-overlooked brominating agents (including BrCl, Br2, BrOCl, Br2O) can form in chlorinated waters, albeit at generally lower concentrations than HOBr. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service