Skip to Content
Merck
All Photos(3)

Key Documents

413364

Sigma-Aldrich

3-(Triethoxysilyl)propyl isocyanate

95%

Synonym(s):

Triethoxy(3-isocyanatopropyl)silane, (3-Isocyanatopropyl)triethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C2H5O)3Si(CH2)3NCO
CAS Number:
Molecular Weight:
247.36
Beilstein:
4404652
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

95%

form

liquid

refractive index

n20/D 1.42 (lit.)

bp

283 °C (lit.)

density

0.999 g/mL at 25 °C (lit.)

functional group

amine
isocyanate

SMILES string

CCO[Si](CCCN=C=O)(OCC)OCC

InChI

1S/C10H21NO4Si/c1-4-13-16(14-5-2,15-6-3)9-7-8-11-10-12/h4-9H2,1-3H3

InChI key

FRGPKMWIYVTFIQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

3-(Triethoxysilyl)propyl isocyanate can be used:
  • As a bifunctional reagent (isocyanate and ethoxysilyl groups) for the immobilization of cellulose derivatives onto silica matrix to generate bonded type of chiral stationary phases (CSPs).
  • To convert 5-amino-1,10-phenanthroline into 5-(N,N-bis-3-(triethoxysilyl)propyl)-ureyl-1,10-phenanthroline, a hydrolysable sol–gel precursor.
  • To synthesize ruthenium bipyridyl tethered porous organosilica(Ru-POS) for organo-photocatalysis.
  • To synthesize nanoparticle-supported oxime palladacycle catalyst for Suzuki-Miyaura cross-coupling reaction.

3-(triethoxysilyl)propyl isocyanate (TEPI) was used as a spacer reagent to prepare cellulose based chiral stationary phases (CSPs). TEPI may be used to modify 2-hydroxynicotinic acid to prepare luminescent hybrid materials.2 Luminescent inorganic-organic hybrid materials were prepared by grafting the isocynate with the ortho aminobenzoic acid derivative. TEPI may also be used to prepare 5-(N,N-bis-3-(triethoxysilyl)propyl)ureyl-1,10- phenanthroline (phen-Si).

Signal Word

Danger

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Eye Dam. 1 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

223.7 °F - closed cup

Flash Point(C)

106.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Magnetic nanoparticles supported oxime palladacycle as a highly efficient and separable catalyst for room temperature Suzuki?Miyaura coupling reaction in aqueous media.
Gholinejad M, et al.
Royal Society of Chemistry Advances, 5(61), 49568-49576 (2015)
A luminescent tris (2-thenoyltrifluoroacetonato) europium (III) complex covalently linked to a 1, 10-phenanthroline-functionalised sol?gel glass.
Journal of Materials Chemistry, 14(2), 191-195 (2004)
Synthesis of covalently bonded cellulose derivative chiral stationary phases with a bifunctional reagent of 3-(triethoxysilyl) propyl isocyanate.
Chen X, et al.
Journal of Chromatography A, 1010(2), 185-194 (2003)
Ruthenium bipyridyl tethered porous organosilica: a versatile, durable and reusable heterogeneous photocatalyst.
Jana A, et al.
Chemical Communications (Cambridge, England), 51(53), 10746-10749 (2015)
Novel luminescent molecular-based hybrid organic?inorganic terbium complex covalently bonded materials via sol?gel process
Inorganic Chemistry, 7(6), 747-750 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service