Skip to Content
Merck
All Photos(2)

Key Documents

468789

Sigma-Aldrich

Dimethylmalononitrile

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C(CN)2
CAS Number:
Molecular Weight:
94.11
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

bp

120 °C/33 mmHg (lit.)

mp

31-33 °C (lit.)

SMILES string

CC(C)(C#N)C#N

InChI

1S/C5H6N2/c1-5(2,3-6)4-7/h1-2H3

InChI key

BCMJJXWXMZYZKN-UHFFFAOYSA-N

Application

Dimethylmalononitrile may be employed as a starting reagent in the synthesis of bisoxazolines and (4′S,5′S)-2,2-bis[4′-hydroxymethyl-5′-phenyl-1′,3′-oxazolin-2′-yl]propane.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

145.4 °F - closed cup

Flash Point(C)

63 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dimethylmalononitrile. Low-temperature heat capacity, vapor pressure, density, and chemical thermodynamics of the crystalline, liquid, and gaseous phases.
Ribner A and Westrum Jr EF.
The Journal of Physical Chemistry, 71(5), 1208-1215 (1967)
Synthesis of non-symmetric bisoxazoline compounds. An easy way to reach tailored chiral ligands.
Garcia JI, et al.
Tetrahedron Asymmetry, 17(15), 2270-2275 (2006)
A new copper(II)/isopropylidene-2,2-bis(oxazoline) catalyst and its stable reactive complex with acryloyloxazolidinone in enantioselective reactions.
Giovanni Desimoni et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 15(38), 9674-9677 (2009-08-18)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service