Skip to Content
Merck
All Photos(2)

Key Documents

450995

Sigma-Aldrich

Manganese(II) chloride

AnhydroBeads, −10 mesh, 99.999% trace metals basis

Synonym(s):

Manganese dichloride, Sacchite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
MnCl2
CAS Number:
Molecular Weight:
125.84
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23

product line

AnhydroBeads

Quality Level

Assay

99.999% trace metals basis

impurities

≤15.0 ppm Trace Metal Analysis

particle size

−10 mesh

mp

652 °C (lit.)

density

2.98 g/mL at 25 °C (lit.)

SMILES string

Cl[Mn]Cl

InChI

1S/2ClH.Mn/h2*1H;/q;;+2/p-2

InChI key

GLFNIEUTAYBVOC-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

A novel high-spin hexanuclear Mn(III) cluster, [NaMn6(OMe)12(dbm)6]+, was synthesized via MnCl2 oxidation with simultaneous NaOMe promoted aggregation. This cluster has an unprecedented cyclic structure and 24 unpaired electrons in its ground state. Also used in the formation of an active Mn(0) species useful for radical cyclization reactions.

Legal Information

AnhydroBeads is a trademark of Sigma-Aldrich Co. LLC

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Dam. 1 - STOT RE 2

Target Organs

Brain

Storage Class Code

6.1D - Non-combustible acute toxic Cat.3 / toxic hazardous materials or hazardous materials causing chronic effects

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Abbati, G.L. et al.
Inorganic Chemistry, 37, 1430-1430 (1998)
Tang, J. et al.
Tetrahedron, 55, 1893-1893 (1999)
M R Sepúlveda et al.
Contrast media & molecular imaging, 7(4), 426-434 (2012-06-01)
Manganese is a vital element and cofactor of many key enzymes, but it is toxic at high levels, causing pronounced disturbances in the mammalian brain. Magnetic resonance imaging (MRI) studies using manganese ions as a paramagnetic contrast agent are often
Asad A Aboud et al.
Neurotoxicology, 33(6), 1443-1449 (2012-10-27)
Manganese (Mn) is an environmental risk factor for Parkinson's disease (PD). Recessive inheritance of PARK2 mutations is strongly associated with early onset PD (EOPD). It is widely assumed that the influence of PD environmental risk factors may be enhanced by
Elizabeth Greally et al.
Journal of cardiovascular magnetic resonance : official journal of the Society for Cardiovascular Magnetic Resonance, 15, 4-4 (2013-01-18)
Manganese-enhanced cardiovascular magnetic resonance (MECMR) can non-invasively assess myocardial calcium influx, and calcium levels are known to be elevated in muscular dystrophy cardiomyopathy based on cellular studies. Left ventricular functional studies and MECMR were performed in mdx mice (model of

Articles

The prevailing strategies for heat and electric-power production that rely on fossil and fission fuels are having a negative impact on the environment and on our living conditions.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service