Skip to Content
Merck
All Photos(1)

Documents

43697

Sigma-Aldrich

rac-1,2-Distearoylglycerol

≥99% (GC)

Synonym(s):

rac-Glycerol 1,2-distearate, DL-1,2-Distearin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C39H76O5
CAS Number:
Molecular Weight:
625.02
Beilstein:
1730305
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥99% (GC)

mp

72-74 °C

SMILES string

CCCCCCCCCCCCCCCCCC(=O)OCC(CO)OC(=O)CCCCCCCCCCCCCCCCC

InChI

1S/C39H76O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-36-37(35-40)44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h37,40H,3-36H2,1-2H3

InChI key

UHUSDOQQWJGJQS-UHFFFAOYSA-N

General description

rac-1,2-Distearoylglycerol is an 1,2-diacylglycerol.

Application

rac-1,2-Distearoylglycerol may be used for the in vitro assay for monoacylglycerol transferase (MGAT) and diacylglycerol acyltransferase (DGAT) activity.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jingsong Cao et al.
The Journal of biological chemistry, 278(16), 13860-13866 (2003-02-11)
Acyl-CoA:monoacylglycerol transferase (MGAT) plays a predominant role in dietary fat absorption in the small intestine, where it catalyzes the first step of triacylglycerol resynthesis in enterocytes for chylomicron formation and secretion. Although the mouse small intestine exhibits the highest MGAT
Jingsong Cao et al.
The Journal of biological chemistry, 278(28), 25657-25663 (2003-05-06)
Acyl-CoA:monoacylglycerol acyltransferase (MGAT) plays an important role in dietary fat absorption by catalyzing a rate-limiting step in the re-synthesis of diacylglycerols in enterocytes. The present study reports further characterization of MGAT2, a newly identified intestinal MGAT (Cao, J., Lockwood, J.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service