Skip to Content
Merck
All Photos(2)

Key Documents

D9135

Sigma-Aldrich

1,2-Dipalmitoyl-sn-glycerol

≥99%

Synonym(s):

(S)-1,2-Dipalmitin, (S)-Glycerol 1,2-dipalmitate, 1,2-Dihexadecanoyl-sn-glycerol, DG(16:0/16:0/0:0), Hexadecanoic acid 1,1′-[(1S)-1-(hydroxymethyl)-1,2-ethanediyl] ester, NSC 269964

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C35H68O5
CAS Number:
Molecular Weight:
568.91
Beilstein:
1730209
EC Number:
MDL number:
UNSPSC Code:
51191904
PubChem Substance ID:
NACRES:
NA.25
Pricing and availability is not currently available.

biological source

Porcine pancreas
bovine skin

Quality Level

Assay

≥99%

form

powder

impurities

1,3-isomer, essentially free

functional group

ester

lipid type

neutral glycerides

shipped in

ambient

storage temp.

−20°C

SMILES string

CCCCCCCCCCCCCCCC(=O)OC[C@H](CO)OC(=O)CCCCCCCCCCCCCCC

InChI

1S/C35H68O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(37)39-32-33(31-36)40-35(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h33,36H,3-32H2,1-2H3/t33-/m0/s1

InChI key

JEJLGIQLPYYGEE-XIFFEERXSA-N

Looking for similar products? Visit Product Comparison Guide

Preparation Note

Prepared by a modification of the procedure of Sowdon, J.C. and Fisher, H.O.l., J. Am. Chem. Soc., 63, 3244 (1941).

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Takashi Watanabe et al.
Journal of lipid research, 58(12), 2334-2347 (2017-10-14)
Thraustochytrids are marine single-cell protists that produce large amounts of PUFAs, such as DHA. They accumulate PUFAs in lipid droplets (LDs), mainly as constituent(s) of triacylglycerol (TG). We identified a novel protein in the LD fraction of
Complexity of Complement Resistance Factors Expressed by
Amaro F Sanchez-Larrayoz et al.
Journal of immunology (Baltimore, Md. : 1950), 199(8), 2803-2814 (2017-09-01)
Luciana Mercedes Pujol-Lereis et al.
Journal of insect physiology, 87, 53-62 (2016-02-13)
The remodeling of membrane composition by changes in phospholipid head groups and fatty acids (FA) degree of unsaturation has been associated with the maintenance of membrane homeostasis under stress conditions. Overall lipid levels and the composition of cuticle lipids also
Michael Zick et al.
Molecular biology of the cell, 24(23), 3746-3753 (2013-10-04)
The fusion of yeast vacuolar membranes depends on the disassembly of cis-soluble N-ethylmaleimide-sensitive factor attachment protein receptor (SNARE) complexes and the subsequent reassembly of new SNARE complexes in trans. The disassembly of cis-SNARE complexes by Sec17/Sec18p releases the soluble SNARE
Chiaki Murakami et al.
Analytical biochemistry, 526, 43-49 (2017-03-21)
Phosphatidylcholine (PC)-specific phospholipase C (PC-PLC) hydrolyzes PC to generate the second messenger 1,2-diacylglycerol (DG) and phosphocholine. PC-PLC plays pivotal roles in inflammation, carcinogenesis, tumor progression, atherogenesis, and subarachnoid hemorrhage. Although the activity of PC-PLC in mammalian tissues was discovered approximately

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service