Skip to Content
Merck
All Photos(3)

Documents

427551

Sigma-Aldrich

Benzophenone

purified by sublimation, ≥99%

Synonym(s):

Diphenyl ketone, Diphenylmethanone, NSC 8077

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H5)2CO
CAS Number:
Molecular Weight:
182.22
Beilstein:
1238185
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

vapor pressure

1 mmHg ( 108 °C)

Assay

≥99%

form

powder or crystals

purified by

sublimation

bp

305 °C (lit.)

mp

47-51 °C (lit.)

SMILES string

O=C(C1=CC=CC=C1)C2=CC=CC=C2

InChI

1S/C13H10O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1-10H

InChI key

RWCCWEUUXYIKHB-UHFFFAOYSA-N

Gene Information

rat ... Ar(24208)

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Benzophenone is an organic compound that can be prepared by the atmospheric oxidation of diphenyl methane with copper naphthenate catalyst. It can also be synthesized by the Friedel-Crafts acylation of benzene with benzoyl chloride.

Application

Benzophenone is mainly used as a photoinitiator molecule in a variety of 3D printing applications. It has a high quantum yield that makes it useful in the development of UV curable photopolymers.

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 1B - STOT RE 2 Oral

Target Organs

Liver,Kidney

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 1

Flash Point(F)

280.4 °F - closed cup

Flash Point(C)

138 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Direct laser writing: Principles and materials for scaffold 3D printing
Selimis A, et al.
Microelectronic Engineering, 132, 83-89 (2015)
Functionalizing micro-3D-printed protein hydrogels for cell adhesion and patterning
Hernandez DS, et al.
Journal of materials chemistry. B, 4(10), 1818-1826 (2016)
3D printing of polymer nanocomposites via stereolithography
Manapat JZ, et al.
Macromolecular Materials and Engineering, 302(9), 1600553-1600553 (2017)
Ketones
Siegel H and Eggersdorfer M
Ullmann's Encyclopedia of Industrial Chemistry (2000)
Claire Chatalova-Sazepin et al.
Angewandte Chemie (International ed. in English), 54(18), 5443-5446 (2015-03-05)
A three-component carboetherification of unactivated alkenes has been developed allowing the rapid building of complexity from simple starting materials. A wide range of α-substituted styrenes underwent smooth reactions with unactivated alkyl nitriles and alcohols to afford γ-alkoxy alkyl nitriles with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service