Skip to Content
Merck
All Photos(1)

Key Documents

427292

Sigma-Aldrich

1-Bromo-2-butyne

≥98%

Synonym(s):

1-(Bromomethyl)-2-methylacetylene, 2-Butyn-1-yl bromide, 2-Butynyl bromide, 4-Bromobut-2-yne

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C≡CCH2Br
CAS Number:
Molecular Weight:
132.99
Beilstein:
605306
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥98%

form

liquid

refractive index

n20/D 1.508 (lit.)

bp

40-41 °C/20 mmHg (lit.)

density

1.519 g/mL at 25 °C (lit.)

functional group

alkyl halide
bromo

SMILES string

CC#CCBr

InChI

1S/C4H5Br/c1-2-3-4-5/h4H2,1H3

InChI key

LNNXOEHOXSYWLD-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Bromo-2-butyne is a propargyl bromide derivative. It is one of the constitutional isomer of bromo butyne. Its Br-loss threshold photoionization breakdown diagram has been analyzed to derive dissociative photoionization thresholds to C4H5+ production. It participates in the preparation of linagliptin.

Application

1-Bromo-2-butyne was used in the alkylation of L-tryptophan methyl ester. It was used as a source to generate CH3CCCH2 radicals to investigate the reaction kinetics of these radicals with NO and NO2.
It may be used in the synthesis of the following:
  • 4-butynyloxybenzene sulfonyl chloride
  • mono-propargylated diene derivative
  • isopropylbut-2-ynylamine
  • allenylcyclobutanol derivatives
  • allyl-[4-(but-2-ynyloxy)phenyl]sulfane
  • allenylindium
  • alkynyl alcohols
  • axially chiral teranyl compounds
Exploited in the synthesis of axially chiral teranyl compounds.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

96.8 °F - closed cup

Flash Point(C)

36 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Exploring the functional space of thiiranes as gelatinase inhibitors using click chemistry.
Testero SA, et al.
ARKIVOC (Gainesville, FL, United States), 7, 221-236 (2011)
Kinetics of resonance stabilized CH3CCCH2 radical reactions with NO and NO2.
Rissanen MP, et al.
Chemical Physics Letters, 543, 28-33 (2012)
Acetylenic TACE inhibitors. Part 1. SAR of the acyclic sulfonamide hydroxamates.
Levin JI, et al.
Bioorganic & Medicinal Chemistry Letters, 13(16), 2799-2803 (2003)
Imaging breakdown diagrams for bromobutyne isomers with photoelectron-photoion coincidence.
Bodi A and Hemberger P.
Physical Chemistry Chemical Physics, 16(2), 505-515 (2014)
Reaction of zirconacyclopentadienes with electrophiles such as benzaldehyde, methyl methacrylate and 1-bromo-2-butyne after treatment with RLi.
Seki T, et al.
Tetrahedron Letters, 45(49), 9041-9043 (2004)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service