Skip to Content
Merck
All Photos(1)

Key Documents

419141

Sigma-Aldrich

Zirconium

foil, thickness 0.1 mm, 99.98% trace metals basis

Synonym(s):

Zirconium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zr
CAS Number:
Molecular Weight:
91.22
EC Number:
MDL number:
UNSPSC Code:
12141751
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

99.98% trace metals basis

form

foil

resistivity

40 μΩ-cm, 20°C

thickness

0.1 mm

bp

4377 °C (lit.)

mp

1852 °C (lit.)

density

6.5 g/mL at 25 °C (lit.)

SMILES string

[Zr]

InChI

1S/Zr

InChI key

QCWXUUIWCKQGHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Quantity

50×150 mm (approximately 4.6 g)

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

The enigma of the bilayered zirconia restoration.
Bernard E Keough et al.
Dentistry today, 32(5), 68-68 (2013-06-01)
Gye-Jeong Oh et al.
Journal of nanoscience and nanotechnology, 13(6), 4252-4255 (2013-07-19)
In this study, the effect of transition metal dopants, originally added as colouring agents, on the mechanical properties and biocompatibility of sintered zirconia was investigated. This study confirmed that transition metal dopants could have a slight detrimental effect on the
An ideal full-arch tooth replacement option: CAD/CAM zirconia screw-retained implant bridge.
Michael Tischler et al.
Dentistry today, 32(5), 98-102 (2013-06-01)
B Kuter et al.
European journal of paediatric dentistry : official journal of European Academy of Paediatric Dentistry, 14(2), 90-94 (2013-06-14)
The objective of this study was to assess the effect of heat application on the mechanical properties of glass ionomer cements. experimental design. The effect of heat on glass ionomer cements during their setting was evaluated by measuring compressive strength
Removing all-ceramic restorations with lasers.
Jeffrey P Cranska
Dentistry today, 32(6), 101-102 (2013-06-28)

Articles

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

Combinatorial Materials Science identifies breakthrough materials through systematic exploration, aiding material discovery.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service