Skip to Content
Merck
All Photos(2)

Documents

403296

Sigma-Aldrich

Zirconium

powder, −100 mesh

Synonym(s):

Zirconium element

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
Zr
CAS Number:
Molecular Weight:
91.22
MDL number:
UNSPSC Code:
12141751
PubChem Substance ID:
NACRES:
NA.23

form

powder

contains

solution 1:10 pentanol to water as stabilizer

resistivity

40 μΩ-cm, 20°C

particle size

−100 mesh

bp

4377 °C (lit.)

mp

1852 °C (lit.)

density

6.5 g/mL at 25 °C (lit.)

SMILES string

[Zr]

InChI

1S/Zr

InChI key

QCWXUUIWCKQGHC-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Starting material for a thermite reaction synthesis of a new compound, ZrXFe1-XO2, which is the first reported example of a compound containing a direct Fe-Zr bond.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Pyr. Sol. 1 - Water-react 1

Supplementary Hazards

Storage Class Code

4.2 - Pyrophoric and self-heating hazardous materials

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Ghigna, P. et al.
Journal of the American Chemical Society, 121, 301-301 (1999)
Thijs H Oude Munnink et al.
Journal of nuclear medicine : official publication, Society of Nuclear Medicine, 54(6), 929-935 (2013-04-30)
Placental growth factor (PlGF) is a member of the proangiogenic vascular endothelial growth factor family, which is upregulated in many tumors. RO5323441, a humanized monoclonal antibody against PlGF, showed antitumor activity in human tumor xenografts. We therefore aimed to radiolabel
The enigma of the bilayered zirconia restoration.
Bernard E Keough et al.
Dentistry today, 32(5), 68-68 (2013-06-01)
O Tymofiyeva et al.
Dento maxillo facial radiology, 42(6), 20120271-20120271 (2013-04-24)
To investigate the potential influence of standard dental materials on dental MRI (dMRI) by estimating the magnetic susceptibility with the help of the MRI-based geometric distortion method and to classify the materials from the standpoint of dMRI. A series of
New CAD/CAM materials and blocks for chairside procedures.
M Zimmermann et al.
International journal of computerized dentistry, 16(2), 173-181 (2013-08-13)

Articles

Mechanical alloying is a “brute force” method of affecting alloying and chemical reactions. The mixture of reactant powders and several balls are placed in the milling jar of a high-energy ball mill, for example, a shaker mill or a planetary mill.

Permanent magnets are an essential technology for energy conversion. Motors and generators are used to convert energy between electrical and mechanical forms.

Biomedical implants are essentially foreign substances within the human body that must survive many years’ exposure to demanding mechanical and physiological conditions. Despite these challenges, metal implants have been widely used to substitute for or rebuild hard tissues such as bones and teeth.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service