Skip to Content
Merck
All Photos(2)

Key Documents

415472

Sigma-Aldrich

2-Indanone oxime

96%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H9NO
CAS Number:
Molecular Weight:
147.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

96%

mp

152-154 °C (lit.)

functional group

oxime

SMILES string

O\N=C1/Cc2ccccc2C1

InChI

1S/C9H9NO/c11-10-9-5-7-3-1-2-4-8(7)6-9/h1-4,11H,5-6H2

InChI key

UJLFLOAXKVGYIA-UHFFFAOYSA-N

General description

Reduction of 2-indanone oxime to 2-aminoindane has been reported.

Application

2-Indanone oxime may be used for the preparation of 2-aminoindane hydrochloride by using active palladium catalyst.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Physiologically active indanamines1.
Levin N, et al.
The Journal of Organic Chemistry, 9(4), 380-391 (1944)
The reduction of 2-indanone oxime to 2-aminoindane. Methods and mechanisms.
Rosen WE and Green MJ.
The Journal of Organic Chemistry, 28(10), 2797-2804 (1963)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service