Skip to Content
Merck
All Photos(1)

Key Documents

A10507

Sigma-Aldrich

Acetone oxime

98%

Synonym(s):

β-Isonitrosopropane, 2-Propanone oxime, Acetoxime, Dimethyl ketoxime, N-Isopropylidenehydroxylamine, Propanone oxime

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2C=NOH
CAS Number:
Molecular Weight:
73.09
Beilstein:
1560146
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.02

Quality Level

Assay

98%

form

crystals

bp

135 °C (lit.)

mp

60-63 °C (lit.)

density

0.901 g/mL at 25 °C (lit.)

SMILES string

C\C(C)=N/O

InChI

1S/C3H7NO/c1-3(2)4-5/h5H,1-2H3

InChI key

PXAJQJMDEXJWFB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Carc. 2 - Eye Dam. 1 - Flam. Sol. 2 - Skin Sens. 1B

Storage Class Code

4.1B - Flammable solid hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M Rysková et al.
Folia biologica, 43(1), 19-24 (1997-01-01)
The genotoxic effects of N-nitroso-N-methylurea (MNU) and acetone oxime (ACOX) were tested in the Somatic Mutation and Recombination Test (SMART) in Drosophila melanogaster. We have performed the same assay on transgenic flies expressing the human gene encoding a glutathione S-transferase
C Kohl et al.
Carcinogenesis, 13(7), 1091-1094 (1992-07-01)
The hepatocarcinogenicity of acetoxime has been tentatively linked with its metabolic oxidation to the potent genotoxicant and carcinogen propane 2-nitronate (P2-N). In order to test the hypothesis that acetoxime is metabolized to P2-N, the oxime (20 mM) was incubated with
Stefanie Zorbas-Seifried et al.
Molecular pharmacology, 71(1), 357-365 (2006-10-20)
The presence of cis-configured exchangeable ligands has long been considered a prerequisite for antitumor activity of platinum complexes, but over the past few years, several examples violating this structure-activity relationship have been recognized. We report here on studies with the
S S Mirvish et al.
Journal of the National Cancer Institute, 69(4), 961-962 (1982-10-01)
Acetoxime was tested for carcinogenicity by chronic administration in the drinking water to male and female outbred MRC-Wistar rats. The dose of 1.0 g/liter was administered 5 days/week for 18 months (total dose, 6.2--7.0 g/rat). The test compound induced benign
N Guo et al.
Carcinogenesis, 11(9), 1659-1662 (1990-09-01)
The hepatocarcinogens 2-nitropropane and acetoxime have previously been found to induce a specific and qualitatively identical pattern of base damage in rat liver DNA and RNA, including the induction of increased levels of 8-hydroxyguanine. Because both 2-nitropropane and acetoxime are

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service