Skip to Content
Merck
All Photos(1)

Key Documents

413046

Sigma-Aldrich

Lithium acetylacetonate

97%

Synonym(s):

2,4-Pentanedione lithium derivative, Acetylacetone lithium derivative, Li(acac)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH=C(OLi)CH3
CAS Number:
Molecular Weight:
106.05
Beilstein:
4575087
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

97%

form

solid

reaction suitability

core: lithium

mp

250 °C (dec.) (lit.)

SMILES string

[Li+].C\C([O-])=C\C(C)=O

InChI

1S/C5H8O2.Li/c1-4(6)3-5(2)7;/h3,6H,1-2H3;/q;+1/p-1/b4-3-;

InChI key

JTEOOCAGEXVCBQ-LNKPDPKZSA-M

Looking for similar products? Visit Product Comparison Guide

General description

Lithium acetylacetonate is a coordination compound utilized as a precursor for producing lithium-containing materials, which finds application in organic light-emitting diodes (OLEDs), lithium-ion batteries, and as a catalyst in organic synthesis. Additionally, it serves as a stabilizer for polymeric electrolytes in lithium batteries and as a curing agent for epoxy resins.

Application

  • Lithium acetylacetonate [Li(acac)] is used in the preparation of LiMn2O4 nanoparticles.
  • Li(acac) covered with aluminium was used as an efficient electron injection layer in research & development of organic light-emitting devices (OLEDs).
  • Fabrication of electrospun graphene-embedded Li4Ti5O12 nanofibers in research & development of lithium-ion batteries has been reported.

Features and Benefits

Its solubility in water enhances its effectiveness as a catalyst for organic reactions.

Pictograms

Health hazardExclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Repr. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Wei Qian et al.
ChemSusChem, 12(6), 1169-1178 (2019-01-09)
Green synthesis of high-molecular-weight isosorbide-based polycarbonate (PIC) with excellent properties is a tremendous challenge and is profoundly influenced by the precursor. Herein, an ecofriendly catalyst was employed to obtain the more reactive PIC precursor dicarboxymethyl isosorbide (DC) with 99.0 % selectivity
Electrochemistry of LiMn 2 O 4 nanoparticles made by flame spray pyrolysis
Patey TJ, et al.
Physical Chemistry Chemical Physics, 11(19), 3756-3761 null
Graphene as a conductive additive to enhance the high-rate capabilities of electrospun Li 4 Ti 5 O 12 for lithium-ion batteries.
Zhu N, et al.
Electrochimica Acta, 55(20), 5813-5818 (2010)
Improved electron injection in organic light-emitting devices with a lithium acetylacetonate [Li (acac)]/aluminium bilayer cathode
Shi S, et al.
Semiconductor Science and Technology, 22(3), 249-249 (2007)
Improved electron injection in organic light-emitting devices with a lithium acetylacetonate [Li (acac)]/aluminium bilayer cathode
Shengwei Shi, et al.
Semiconductor Science and Technology, 22, 249-249 (2007)

Articles

Few Monolayer Atomic Layer Deposition (ALD) on Surfaces and Interfaces for Energy Applications

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service