Skip to Content
Merck
All Photos(1)

Key Documents

397237

Sigma-Aldrich

Acetanilide

zone-refined, purified by sublimation, ≥99.95%

Synonym(s):

N-Phenylacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CONHC6H5
CAS Number:
Molecular Weight:
135.16
Beilstein:
606468
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39031308
PubChem Substance ID:
NACRES:
NA.22

vapor density

4.65 (vs air)

Quality Level

vapor pressure

1 mmHg ( 114 °C)

Assay

≥99.95%

form

solid

autoignition temp.

1004 °F

purified by

sublimation

bp

304 °C (lit.)

mp

113-115 °C (lit.)

functional group

amide

SMILES string

CC(=O)Nc1ccccc1

InChI

1S/C8H9NO/c1-7(10)9-8-5-3-2-4-6-8/h2-6H,1H3,(H,9,10)

InChI key

FZERHIULMFGESH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Acetanilide is an aniline derivative. Crystal structure of acetanillide has been studied by X-ray diffraction. It crystallizes in orthorhombic system having space group of Pbca. Its photolysis has been studied in the presence of β-cyclodextrin (cycloheptaamylose). Aminoacetophenone derivatives are obtained by the Fries rearrangement of acetanilide in the presence zeolites Y and β.

Application

Acetanilide may be used in the synthesis of ortho-haloacetanilides via regioselective C-H functionalization/halogenation reaction in the presence of Pd(OAc)2 and Cu(OAc)2 catalyst. It may be used in the preparation of chlorosubstituted acetanilides.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

321.8 °F - closed cup

Flash Point(C)

161 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oxidative Chlorination of Acetanilides by Metal Chlorides-Hydrogen Peroxide in Acid-Aqueous Medium Systems.
Jerzy G and Slawomir G.
Synthetic Communications, 27(18), 3291-3299 (1997)
A REMPI and ZEKE Spectroscopic Study of trans-Acetanilide? H2O and Comparison to Ab Initio CASSCF Calculations.
Ullrich S and Muller-Dethlefs K.
The Journal of Physical Chemistry A, 106(40), 9188-9195 (2002)
Xiaobing Wan et al.
Journal of the American Chemical Society, 128(23), 7416-7417 (2006-06-08)
Highly regioselective C-H functionalization/halogenation of acetanilides to produce ortho-haloacetanilides was catalyzed by Pd(OAc)2 and Cu(OAc) 2 with CuX2 as the halogen source.
The crystal structure of acetanilide.
Brown CJ and Corbridge DEC.
Acta Crystallographica, 7(11), 711-715 (1954)
Fries rearrangement of acetanilide over zeolite catalysts.
Balkus Jr KJ, et al.
J. Mol. Catal. A: Chem., 134(1), 137-143 (1998)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service