Skip to Content
Merck
All Photos(1)

Key Documents

38510

Sigma-Aldrich

Dimercaptomaleonitrile disodium salt hydrate

≥95.0% (calc. based on dry substance, NT)

Synonym(s):

(Z)-2,3-Dimercapto-2-butenedinitrile disodium salt, Bahr′s salt, Disodium (Z)-1,2-dicyano-1,2-ethylenedithiolate, cis-1,2-Dicyano-1,2-dimercaptoethylene disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4N2Na2S2 · xH2O
CAS Number:
Molecular Weight:
186.17 (anhydrous basis)
Beilstein:
3725169
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

≥95.0% (calc. based on dry substance, NT)

concentration

~10% (crystal water)

functional group

nitrile

SMILES string

O.[Na]S\C(C#N)=C(/S[Na])C#N

InChI

1S/C4H2N2S2.2Na.H2O/c5-1-3(7)4(8)2-6;;;/h7-8H;;;1H2/q;2*+1;/p-2/b4-3-;;;

InChI key

DWYJCSXARKJSBC-FGSKAQBVSA-L

General description

Dimercaptomaleonitrile disodium salt is commonly employed as ligand for transition metals.

Application

Dimercaptomaleonitrile disodium salt may be used in the preparation of cis-2,3-bis[6-(1,2-closo-dodecarboranyl)hexylthio] maleonitrile.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

P.I. Clemenson
Coordination Chemistry Reviews, 106, 171-171 (1990)
Anna Salvati et al.
Biophysical chemistry, 131(1-3), 43-51 (2007-10-06)
The synthesis, characterization and liposome insertion of a novel magnesium(II) carboranyl-porphyrazine, i.e. [2,3,7,8,12,13,17,18-octakis-(1,2-dicarba-closo-dodecaboranyl)-hexylthio-5,10,15,20-porphyrazine]magnesium(II) complex, MgHECSPz, are described. MgHECSPz was designed to improve the potentiality in multiple approach anticancer therapy. Liposomal formulations with different surface charge were prepared as delivering agents.
Fritz Sieber et al.
Phosphorus, sulfur, and silicon and the related elements, 180(3-4), 647-657 (2006-02-24)
Elemental selenium generated by the photobleaching of selenomerocyanine dyes forms conjugates with serum albumin and serum lipoproteins that are toxic to leukemia and selected solid tumor cells but well tolerated by normal CD34-positive hematopoietic stem and progenitor cells. Serum albumin

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service