Skip to Content
Merck
All Photos(2)

Key Documents

367877

Sigma-Aldrich

Potassium carbonate

99.995% trace metals basis

Synonym(s):

Dipotassium carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
K2CO3
CAS Number:
Molecular Weight:
138.21
Beilstein:
4267587
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder and chunks
solubility:
H2O: soluble 138 g/L at 20 °C (completely)
Pricing and availability is not currently available.

Quality Level

Assay

99.995% trace metals basis

form

powder and chunks

impurities

≤55.0 ppm Trace Metal Analysis

mp

891 °C (lit.)

solubility

H2O: soluble 138 g/L at 20 °C (completely)

SMILES string

[K+].[K+].[O-]C([O-])=O

InChI

1S/CH2O3.2K/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

BWHMMNNQKKPAPP-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

General description

Potassium carbonate (K2CO3) is a water-soluble inorganic compound that is primarily derived from potash. It is industrially prepared by reacting potassium hydroxide and carbon dioxide. The solution crystallizes to form potash hydrate, which can further be heated above 200°C to produce the anhydrous salt.[1][2]

Application

K2CO3 can be used for a variety of applications such as:
  • production of soaps and glasses[3][3]
  • drying agent for organic based compounds like ketones, alcohols, and amines[4]
  • buffering agent to produce wine[5]
  • ingredient in welding and other coatings[5]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Potassium compounds
Schultz H, et al.
Ullmann's Encyclopedia of Industrial Chemistry (2000)
A Convenient Stereoselective Reduction of gem-Dibromides with a Combination of Dimethyl Phosphite and Potassium Carbonate
Zhao Y, et al.
Phosph. Sulfur Relat. Elem. (2015)
Formic acid decomposition over palladium based catalysts doped by potassium carbonate
Jia L, et al.
Catalysis Today (2015)
A study of the vapour-liquid equilibrium of CO2 in mixed solutions of potassium carbonate and potassium glycinate
Lee A, et al.
International Journal of Greenhouse Gas Control, 36, 27-33 (2015)
Highly Selective and Sensitive Voltammetric Sensor Based on Ruthenium Nanoparticle Anchored Calix [4] amidocrown-5 Functionalized Reduced Graphene Oxide: Simultaneous Determination of Quercetin, Morin and Rutin in Grape Wine
Elcin S, et al.
Electroanalysis, 28(3), 611-619 (2016)

Questions

1–2 of 2 Questions  
  1. How is shipping temperature determined? And how is it related to the product storage temperature?

    1 answer
    1. Products may be shipped at a different temperature than the recommended long-term storage temperature. If the product quality is sensitive to short-term exposure to conditions other than the recommended long-term storage, it will be shipped on wet or dry-ice. If the product quality is NOT affected by short-term exposure to conditions other than the recommended long-term storage, it will be shipped at ambient temperature. As shipping routes are configured for minimum transit times, shipping at ambient temperature helps control shipping costs for our customers. For more information, please refer to the Storage and Transport Conditions document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/316/622/storage-transport-conditions-mk.pdf

      Helpful?

  2. How can I determine the shelf life / expiration / retest date of this product?

    1 answer
    1. If this product has an expiration or retest date, it will be shown on the Certificate of Analysis (COA, CofA). If there is no retest or expiration date listed on the product's COA, we do not have suitable stability data to determine a shelf life. For these products, the only date on the COA will be the release date; a retest, expiration, or use-by-date will not be displayed.
      For all products, we recommend handling per defined conditions as printed in our product literature and website product descriptions. We recommend that products should be routinely inspected by customers to ensure they perform as expected.
      For products without retest or expiration dates, our standard warranty of 1 year from the date of shipment is applicable.
      For more information, please refer to the Product Dating Information document: https://www.sigmaaldrich.com/deepweb/assets/sigmaaldrich/marketing/global/documents/449/386/product-dating-information-mk.pdf

      Helpful?

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service