366773
Zirconium(IV) sulfate hydrate
99.99% trace metals basis
Sign Into View Organizational & Contract Pricing
All Photos(3)
About This Item
Linear Formula:
Zr(SO4)2·xH2O
CAS Number:
Molecular Weight:
283.35 (anhydrous basis)
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
Assay:
99.99% trace metals basis
form:
solid
Recommended Products
Quality Level
Assay
99.99% trace metals basis
form
solid
impurities
≤150.0 ppm Trace Metal Analysis
pH
1-2 (25 °C, 5%)
application(s)
battery manufacturing
SMILES string
O.[Zr+4].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
InChI
1S/2H2O4S.H2O.Zr/c2*1-5(2,3)4;;/h2*(H2,1,2,3,4);1H2;/q;;;+4/p-4
InChI key
UXRMCZYAVOHQNB-UHFFFAOYSA-J
Looking for similar products? Visit Product Comparison Guide
Related Categories
Other Notes
may contain excess sulfuric acid
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service