Skip to Content
Merck
All Photos(2)

Key Documents

328766

Sigma-Aldrich

N-Methyl-N-phenylcarbamoyl chloride

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5N(CH3)COCl
CAS Number:
Molecular Weight:
169.61
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

bp

280 °C (lit.)

mp

87-90 °C (lit.)

storage temp.

2-8°C

SMILES string

CN(C(Cl)=O)c1ccccc1

InChI

1S/C8H8ClNO/c1-10(8(9)11)7-5-3-2-4-6-7/h2-6H,1H3

InChI key

CPGWSLFYXMRNDV-UHFFFAOYSA-N

General description

Solvolyses of N-methyl-N-phenylcarbamoyl chlorides in aqueous binary mixtures of acetone, ethanol, methanol and in water, D2O and 50% D2O-CH3OD were investigated. Iridium-catalyzed annulation of N-methyl-N-phenylcarbamoyl chloride with internal alkyne was reported.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tomohiro Iwai et al.
Journal of the American Chemical Society, 132(28), 9602-9603 (2010-06-30)
An iridium complex successfully catalyzed the annulation of various N-arylcarbamoyl chlorides with internal alkynes to afford 2-quinolones in good to excellent yields. The present reaction is widely applicable to substrates with various functionalities. An amide-iridacycle complex was isolated, and it
Solvolyses of N-Methyl-N-phenylcarbamoyl Chlorides with Electron Acceptor Substituents in Aqueous Binary Mixtures.
Koo IS, et al
Bull. Korean Chem. Soc., 22(8), 842-846 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service