Skip to Content
Merck
All Photos(3)

Documents

348015

Sigma-Aldrich

2-Aminobenzylamine

98%

Synonym(s):

(2-Aminomethylphenyl)amine, 2-(Aminomethyl)aniline, 2-(Aminomethyl)benzenamine, 2-Amino-1-benzylamine, [(2-Aminophenyl)methyl]amine, o-Aminobenzylamine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC6H4CH2NH2
CAS Number:
Molecular Weight:
122.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

58-61 °C (lit.)

SMILES string

NCc1ccccc1N

InChI

1S/C7H10N2/c8-5-6-3-1-2-4-7(6)9/h1-4H,5,8-9H2

InChI key

GVOYKJPMUUJXBS-UHFFFAOYSA-N

General description

2-Aminobenzylamine undergoes three-component cyclisation reactions with methyl 3,3,3-trifluoropyruvate, 2-aminobenzylamine and oxo compounds to afford regio- and stereoisomers of tetrahydropyrroloquinazolinones.

Application

2-Aminobenzylamine may be used:
  • in the synthesis of 1,2,3,4-tetrahydroquinazoline oxime, via condensation reaction with 2-(naphthalen-2-yl)-2-oxoacetaldehyde oxime
  • in the synthesis of alkyl 5H-1,4-benzodiazepine-3-carboxylates
  • to modify the phosphate groups on phosphoserine peptides

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Sirikçi et al.
Acta chimica Slovenica, 59(4), 904-911 (2013-09-26)
A novel 1,2,3,4-tetrahydroquinazoline oxime was synthesised from a condensation reaction of 2-(naphthalen-2-yl)-2-oxoacetaldehyde oxime with 2-aminobenzylamine. Subsequently, a-imine oxime complexes of this compound that formed with Co(III) and Ni(II) metal ions were obtained. All structures were characterised by spectral methods (FT-IR
Wen-Yun Hsueh et al.
Journal of medicinal chemistry, 64(3), 1435-1453 (2021-01-26)
In this paper, we present a copper(I)-catalyzed nitrile-addition/N-arylation ring-closure cascade for the synthesis of 5,11-dihydro-6H-indolo[3,2-c]quinolin-6-ones from 2-(2-bromophenyl)-N-(2-cyanophenyl)acetamides. Using CuBr and t-BuONa in dimethylformamide (DMF) as the optimal reaction conditions, the cascade reaction gave the target products, in high yields, with
Lukas K Filak et al.
Organometallics, 32(3), 903-914 (2013-02-23)
Six novel ruthenium(II)- and osmium(II)-arene complexes with three modified indolo[3,2-c]quinolines have been synthesized in situ starting from 2-aminoindoloquinolines and 2-pyridinecarboxaldehyde in the presence of [M(p-cymene)Cl(2)](2) (M = Ru, Os) in ethanol. All complexes have been characterized by elemental analysis, spectroscopic
Lukas K Filak et al.
Organometallics, 30(2), 273-283 (2011-01-22)
The synthesis of new modified indolo[3,2-c]quinoline ligands L(1)-L(8) with metal-binding sites is reported. By coordination to ruthenium- and osmium-arene moieties 16 complexes of the type [(η(6)-p-cymene)M(L)Cl]Cl (1a,b-8a,b), where M is Ru(II) or Os(II) and L is L(1)-L(8), have been prepared.
Orazio A Attanasi et al.
The Journal of organic chemistry, 76(20), 8320-8328 (2011-09-13)
A novel and simple one-pot synthesis of 3-substituted 2,5,6,7-tetrahydro-1H-1,4-diazepin-2-ones from 1,2-diaza-1,3-dienes (DDs) and N-unsubstituted aliphatic 1,3-diamines is described. Here we also report a procedure to selectively obtain alkyl 5H-1,4-benzodiazepine-3-carboxylates from the DDs and 2-aminobenzylamine. Both processes occur by means of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service