Skip to Content
Merck
All Photos(3)

Key Documents

344508

Sigma-Aldrich

Cyclopentene

96%

Synonym(s):

2,2-Dimethylacetyl chloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H8
CAS Number:
Molecular Weight:
68.12
Beilstein:
635707
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

vapor pressure

20.89 psi ( 55 °C)
6.11 psi ( 20 °C)

Assay

96%

form

liquid

autoignition temp.

743 °F

refractive index

n20/D 1.421 (lit.)

bp

44-46 °C (lit.)

mp

−135 °C (lit.)

solubility

water: soluble 0.535 g/L at 25 °C

density

0.771 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

C1CC=CC1

InChI

1S/C5H8/c1-2-4-5-3-1/h1-2H,3-5H2

InChI key

LPIQUOYDBNQMRZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Cyclopentene is a cyclo-olefin.[1] Neopentyl phosphine ligand catalyzed Heck coupling of cyclopentene has been reported.[2] Mechanism of reaction of ground state oxygen atom with cyclopentene has been investigated.[3] Homopolymerization of cyclopentene has been reported.[1] Photocatalytic oxidation of cyclopentene over various titanium(IV) oxide catalyst has been reported.[4]

Application

Cyclopentene was used to investigate the [2+2] cycloaddition of diamond (001) surfaces with alkene.[5]

recommended

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Oral - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

-29.9 °F - closed cup

Flash Point(C)

-34.4 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stereospecific homopolymerization of cyclopentene.
Natta G, et al.
Angewandte Chemie (International Edition in English), 3(11), 723-729 (1964)
Hongmei Zhao et al.
Journal of molecular graphics & modelling, 51, 184-192 (2014-06-18)
The reaction mechanism of the ground state oxygen atom O((3)P) with cyclopentene is investigated theoretically. The triplet and singlet potential energy surfaces are calculated at the CCSD(T)//MP2/6-311G(d,p) level and the minimum energy crossing points (MECPs) between the two surfaces are
Partial photocatalytic oxidation of cyclopentene over titanium (IV) oxide.
Kluson P, et al.
J. Mol. Catal. A: Chem., 242(1), 62-67 (2005)
Cycloaddition chemistry at surfaces: reaction of alkenes with the diamond (001)-2? 1 surface.
Hovis JS, et al.
Journal of the American Chemical Society, 122(4), 732-733 (2000)
Matthew G Lauer et al.
The Journal of organic chemistry, 79(22), 10837-10848 (2014-10-22)
The use of neopentyl phosphine ligands was examined in the coupling of aryl bromides with alkenes. Di-tert-butylneopentylphosphine (DTBNpP) was found to promote Heck couplings with aryl bromides at ambient temperature. In the Heck coupling of cyclic alkenes, the degree of

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service