Skip to Content
Merck
All Photos(1)

Documents

343862

Sigma-Aldrich

1-Bromoheptadecafluorooctane

99%

Synonym(s):

Heptadecafluorooctyl bromide, Perfluorooctyl bromide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)7Br
CAS Number:
Molecular Weight:
498.96
Beilstein:
2016022
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.305 (lit.)

bp

142 °C (lit.)

density

1.93 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)Br

InChI

1S/C8BrF17/c9-7(22,23)5(18,19)3(14,15)1(10,11)2(12,13)4(16,17)6(20,21)8(24,25)26

InChI key

WTWWXOGTJWMJHI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

1-Bromoheptadecafluorooctane (perfluorooctyl bromide) is a perfluorocarbon. Poly(ethylene glycol)ylated polyester nanocapsules of perfluorooctyl bromide were surface-decorated with a RGD (arginine-glycine-aspartic acid) peptide, were used for tumor imaging. Nanocapsules of perfluorooctyl bromide have been developed as dual contrast agent for ultrasonography and Magnetic Resonance Imaging (MRI).

Application

1-Bromoheptadecafluorooctane (perfluorooctyl bromide) can be used as contrast agent in the vascular system for fluorine-19 magnetic resonance imaging or as synthetic oxygen carrier. It was also employed as oxygen carrier in the emulsions used as blood substitutes.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Céline Giraudeau et al.
Angiogenesis, 16(1), 171-179 (2012-10-12)
Molecular imaging with magnetic resonance imaging (MRI) targeted contrast agents has emerged as a promising diagnostic approach in cancer research to detect associated biomarkers. In this work, the potential of (19)F MRI was investigated to detect angiogenesis with α(ν)β(3)-targeted perfluorooctylbromide
G Srinivas Rao et al.
Journal of environmental science & engineering, 52(2), 137-146 (2010-12-01)
A systematic study has been carried out to explore the physico-chemical characteristics of groundwater in Greater Visakhapatnam Municipal Corporation (GVMC) area. Water samples from 32 bore wells, 16 open wells and two reservoirs at various locations in GVMC area were
S A Snyder et al.
Veterinary and comparative oncology, 6(4), 213-223 (2009-01-31)
Human oncology has clearly demonstrated the existence of hypoxic tumours and the problematic nature of those tumours. Hypoxia is a significant problem in the treatment of all types of solid tumours and a common reason for treatment failure. Hypoxia is
Mitsuhiro Isaka et al.
Annals of thoracic and cardiovascular surgery : official journal of the Association of Thoracic and Cardiovascular Surgeons of Asia, 14(6), 363-368 (2009-01-10)
Perfluoro-octyl bromide (PFOB), one of the perfluorochemical oxygen transporters, improved postischemic cardiac dysfunctions. Also norepinephrine (NE) is one of the important inducible factors on reperfusion arrhythmias (ventricular fibrillation [VF]). We used these methods to evaluate the relationship between PFOB emulsion
Ao Li et al.
Acta radiologica (Stockholm, Sweden : 1987), 54(3), 278-283 (2013-01-16)
Multimodal contrast agents usually containing two or more signal components can be used for several imaging modalities. Thus, complimentary information can be obtained by using just one kind of contrast agents. To investigate the potential of superparamagnetic perfluorooctylbromide nanoparticles (M-PFOBNP)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service