Skip to Content
Merck
All Photos(1)

Documents

262145

Sigma-Aldrich

Perfluorohexyl iodide

99%

Synonym(s):

Tridecafluoro-1-iodohexane, 1-Iodoperfluorohexane, 1-Iodotridecafluorohexane, Tridecafluorohexyl iodide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CF3(CF2)5I
CAS Number:
Molecular Weight:
445.95
Beilstein:
1802118
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

liquid

refractive index

n20/D 1.329 (lit.)

bp

117 °C (lit.)

density

2.063 g/mL at 25 °C (lit.)

SMILES string

FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)I

InChI

1S/C6F13I/c7-1(8,3(11,12)5(15,16)17)2(9,10)4(13,14)6(18,19)20

InChI key

BULLJMKUVKYZDJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Perfluorohexyl iodide forms 1:1 complexes with a variety of hydrogen-bond acceptors. It has been grafted onto the isocitronellene/propene copolymer by radical reaction yielding a poly(α-olefin) with fluorinated side chains. The radical addition of perfluorohexyl iodide to vinyl acetate, using azo-bis( isobutyronitrile) (AIBN) as an initiator, has been investigated.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

239.0 °F - closed cup

Flash Point(C)

115 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexander A Marchione et al.
Journal of magnetic resonance (San Diego, Calif. : 1997), 286, 143-147 (2017-12-16)
A train of BURBOP universal rotation pulses has been used to generate a spin lock sufficient to observe TOCSY correlations over a 46 kHz
Zirconocene-MAO-catalyzed homo-and copolymerizations of linear asymmetrically substituted dienes with propene: A novel strategy to functional (co) poly (a-olefin) s.
Hackmann M, et al.
Macromolecular Chemistry and Physics, 199(8), 1511-1517 (1998)
Rafel Cabot et al.
Chemical communications (Cambridge, England), (15)(15), 2005-2007 (2009-04-01)
Quantitative studies of the 1 : 1 complexes formed between perfluorohexyl iodide and a variety of hydrogen-bond acceptors have been used to probe the relationship between halogen bonding, hydrogen bonding, desolvation and the electrostatics of non-covalent interactions.
Effect of reaction conditions on the radical addition of perfluorohexyl iodide to vinyl acetate.
Napoli M, et al.
Journal of Fluorine Chemistry, 57(1), 219-227 (1992)
Wenting Song et al.
Talanta, 175, 413-420 (2017-08-27)
Contaminants of emerging concern are continuously increasing, which makes it important to develop high throughput screening techniques for the evaluation of their potential biological effects, especially endocrine disrupting effects, which would directly influence the population dynamics in environment. A novel

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service