Skip to Content
Merck
All Photos(1)

Key Documents

326739

Sigma-Aldrich

Hexamethyldisiloxane

NMR grade, ≥99.5%

Synonym(s):

HMDSO

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiOSi(CH3)3
CAS Number:
Molecular Weight:
162.38
Beilstein:
1736258
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

≥99.5%

form

liquid

technique(s)

NMR: suitable

refractive index

n20/D 1.377 (lit.)

bp

101 °C (lit.)

mp

−59 °C (lit.)

density

0.764 g/mL at 20 °C (lit.)

SMILES string

C[Si](C)(C)O[Si](C)(C)C

InChI

1S/C6H18OSi2/c1-8(2,3)7-9(4,5)6/h1-6H3

InChI key

UQEAIHBTYFGYIE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameEnvironment

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

21.2 °F - closed cup

Flash Point(C)

-6 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Stavros Beteinakis et al.
Molecules (Basel, Switzerland), 25(15) (2020-07-29)
Edible olive drupes (from Olea europaea L.) are a high-value food commodity with an increasing production trend over the past two decades. In an attempt to prevent fraud issues and ensure quality, the International Olive Council (IOC) issued guidelines for
Soham D Parikh et al.
Materials science & engineering. C, Materials for biological applications, 108, 110345-110345 (2020-01-12)
Carbon Nanotubes (CNTs) are known for effective adhesion, growth, and differentiation of bone, muscle, and cardiac cells. CNTs can provide excellent mechanical and electrical properties for cell scaffolding; however, loose CNTs can cause in-vivo toxicity. To suppress this risk, our
Vikram D Kodibagkar et al.
Frontiers in bioscience : a journal and virtual library, 13, 1371-1384 (2007-11-06)
Over the years many techniques have been devised for the measurement of tissue oxygenation (oximetry). Oximetry using polarographic needle electrodes has long been considered a gold standard. Nuclear Magnetic Resonance (NMR) based oximetry uses exogenously administered reporter molecules such as
M S Yildirim et al.
Journal of oral rehabilitation, 33(10), 775-783 (2006-08-30)
It has been suggested that altering the surface properties of acrylic resin material may change the nature of the adsorbed pellicle affecting denture retention and microbial adherence. This study aimed at evaluating the adsorption of salivary high molecular-weight mucins, a
Andrew Michelmore et al.
Langmuir : the ACS journal of surfaces and colloids, 27(19), 11943-11950 (2011-08-26)
New data shed light on the mechanisms of film growth from low power, low pressure plasmas of organic compounds. These data rebalance the widely held view that plasma polymer formation is due to radical/neutral reactions only and that ions play

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service