Skip to Content
Merck
All Photos(1)

Documents

235679

Sigma-Aldrich

Decamethyltetrasiloxane

97%

Synonym(s):

1,1,1,3,3,5,5,7,7,7-Decamethyltetrasiloxane, 1,1,3,3-Tetramethyl-1,3-bis(trimethylsiloxy)disiloxane, [Dimethyl(trimethylsilyloxy)silyl]oxy-dimethyl-trimethylsilyloxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3SiO[Si(CH3)2O]2Si(CH3)3
CAS Number:
Molecular Weight:
310.69
Beilstein:
1776881
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

vapor density

>1 (vs air)

Assay

97%

form

liquid

refractive index

n20/D 1.389 (lit.)

bp

194 °C (lit.)

mp

−68 °C (lit.)

density

0.854 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)O[Si](C)(C)O[Si](C)(C)O[Si](C)(C)C

InChI

1S/C10H30O3Si4/c1-14(2,3)11-16(7,8)13-17(9,10)12-15(4,5)6/h1-10H3

InChI key

YFCGDEUVHLPRCZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ken-Ichi Shinohara et al.
Scientific reports, 8(1), 13982-13982 (2018-09-20)
As a composite of hybrid organic-inorganic materials, blending hydrophilic silica microparticles with oil-extended rubber can improve vehicle tire performance but the nanometer scale effects of microparticle inclusion have not been thoroughly studied. Here, we used atomic force microscopy (AFM) video
Jiali Cheng et al.
Environmental science and pollution research international, 28(13), 16054-16064 (2020-11-28)
Methyl siloxanes are becoming increasingly prevalent in the environment because of their extensive use in various consumer products. Little is known about the distribution of these chemicals around factories or their presence in road dust. We conducted a survey of
Tri Manh Tran et al.
Chemosphere, 184, 1117-1124 (2017-07-05)
Earlier studies have reported the occurrence of cyclic and linear siloxanes in personal care and household products. Nevertheless, there is a lack of information on the occurrence of siloxanes in indoor air. In this study, four cyclic and six linear
De-Gao Wang et al.
The Science of the total environment, 574, 1254-1260 (2016-09-25)
We measured the concentrations of linear and cyclic volatile methylsiloxanes (VMS) concentrations in the blood plasma of turtles, cormorants, and seals collected from Canadian freshwater and marine ecosystems. A modified quick, easy, cheap, effective, rugged, and safe (QuEChERS) method was
Paul T J Scheepers et al.
Journal of occupational medicine and toxicology (London, England), 12, 33-33 (2017-12-14)
In most reported cases of lung trauma with water proofing products, volatile organic compounds (VOC) have a prominent role. Here we report on a case involving ten workers exposed to a sprayed product containing nanoparticles in a water solution with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service