Skip to Content
Merck
All Photos(1)

Key Documents

324515

Sigma-Aldrich

trans-1,4-Dichloro-2-butene

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClCH2CH=CHCH2Cl
CAS Number:
Molecular Weight:
125.00
Beilstein:
1719693
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

10 mmHg ( 20 °C)

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.488 (lit.)

bp

74-76 °C/40 mmHg (lit.)

mp

1-3 °C (lit.)

density

1.183 g/mL at 25 °C (lit.)

functional group

chloro

storage temp.

2-8°C

SMILES string

ClC\C=C\CCl

InChI

1S/C4H6Cl2/c5-3-1-2-4-6/h1-2H,3-4H2/b2-1+

InChI key

FQDIANVAWVHZIR-OWOJBTEDSA-N

Looking for similar products? Visit Product Comparison Guide

Application

trans-1,4-Dichloro-2-butene may be used for the alkylation of adenine to 9-alkylpurines. It can react with diethyl malonate to form vinylcyclopropane derivative. It can also undergo asymmetric allylic alkylation with Grignard reagents in the presence of copper thiophene carboxylate catalyst.

Other Notes

85%, remainder predominantly cis isomer

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Eye Dam. 1 - Flam. Liq. 3 - Skin Corr. 1B

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

127.4 °F - closed cup

Flash Point(C)

53 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

High Diversity on Simple Substrates: 1, 4?Dihalo?2?butenes and Other Difunctionalized Allylic Halides for Copper?Catalyzed SN2? Reactions.
Falciola C A, et al.
Chemistry?A European Journal , 14(34), 10615-10627 (2008)
Improved selectivity in the preparation of some 1, 1-difunctionalized 3-cyclopentenes. High yield synthesis of 3-cyclopentenecarboxylic acid.
Depres JP and Greene AE.
The Journal of Organic Chemistry, 49(5), 928-931 (1984)
S Phadtare et al.
Journal of medicinal chemistry, 30(2), 437-440 (1987-02-01)
Alkylation of adenine (5a) or 2-amino-6-chloropurine (5b) with excess trans-1,4-dichloro-2-butene (4), effected by K2CO3 in dimethyl sulfoxide or tetra-n-butylammonium fluoride in tetrahydrofuran, led in 90-95% regioselectivity to 9-alkylpurines 6a and 6b. The title compounds 2a and 2b were obtained by
L S Mullin et al.
Drug and chemical toxicology, 23(3), 403-417 (2000-08-26)
This study was conducted to elucidate the time- and dose-response relationships of long-term, low-level 1,4-dichlorobutene-2 (DCB) inhalation exposure to nasal tumor induction in rats. Male Crl:CD BR rats were exposed 6 hours per day, 5 days week to 0, 0.1
S Phadtare et al.
Nucleic acids symposium series, (18)(18), 25-28 (1987-01-01)
Reaction of adenine (1a) or cytosine (1b) with excess 1,4-dichloro-2-butyne catalyzed by K2CO3 in (CH3)2SO gave the 4-chloro-2-butynyl derivatives 2a and 2b. The latter were converted to the 4-hydroxy-2-butynyl compounds 3a and 3b by refluxing in 0.1 M HCl. Isomerization

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service