Skip to Content
AL
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Water Purification
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
trans
-1,4-Dichloro-2-butene
Linear Formula:
ClCH
2
CH=CHCH
2
Cl
CAS Number:
110-57-6
Molecular Weight:
125.00
EC Number:
203-779-7
Products
Product Category
building blocks (2)
Brand
Sigma-Aldrich (2)
Boiling Point (°C)
Formula Weight
Melting Point (°C)
Physical Form
Purity
Quality Segment
Functional Group
Markush Class
Markush Group
Special Grade
Technical (1)
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
324515
trans
-1,4-Dichloro-2-butene,
98%
Expand
View Pricing
215082
trans
-1,4-Dichloro-2-butene,
technical grade, 85%, remainder predominantly
cis
isomer
Expand
View Pricing