Skip to Content
Merck
All Photos(2)

Documents

323578

Sigma-Aldrich

2,2′-Dihydroxy-4-methoxybenzophenone

98%

Synonym(s):

Dioxybenzone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3OC6H3(OH)COC6H4OH
CAS Number:
Molecular Weight:
244.24
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Quality Level

Assay

98%

form

powder

bp

170-175 °C/1 mmHg (lit.)

mp

73-75 °C (lit.)

SMILES string

COc1ccc(c(O)c1)C(=O)c2ccccc2O

InChI

1S/C14H12O4/c1-18-9-6-7-11(13(16)8-9)14(17)10-4-2-3-5-12(10)15/h2-8,15-16H,1H3

InChI key

MEZZCSHVIGVWFI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Wen Luo et al.
Journal of enzyme inhibition and medicinal chemistry, 26(5), 706-711 (2011-01-22)
A novel series of N,N'-bis-methylenedioxybenzyl-alkylenediamines 5a-5g have been designed, synthesized and evaluated as bivalent anti-Alzheimer's disease ligands. The enzyme inhibition assay results indicated that compounds 5e-5g inhibit both acetylcholinesterase (AChE) and butyrylcholinesterase (BuChE) in the micromolar range (IC(50), 2.76-4.24 µM
Vaughn F Sherwood et al.
Cutaneous and ocular toxicology, 31(4), 273-279 (2012-01-20)
Sunscreens are widely utilized due to the adverse effects of ultraviolet (UV) radiation on human health. The safety of their active ingredients as well as that of any modified versions generated during use is thus of concern. Chlorine is used
Stephen E Duirk et al.
Water research, 47(2), 579-587 (2012-11-22)
The transformation of two benzophenone UV filters (Oxybenzone and Dioxybenzone) was examined over the pH range 6-11 in the presence of excess aqueous chlorine. Under these conditions, both UV filters were rapidly transformed by aqueous chlorine just above circumneutral pH
N Negreira et al.
Analytica chimica acta, 638(1), 36-44 (2009-03-21)
A sensitive procedure for the determination of three UV filters: ethylhexyl salicylate (EHS), 3,3,5-trimethylcyclohexyl salicylate (Homosalate, HMS), 2-hydroxy-4-methoxybenzophenone (BP-3) and two related hydroxylated benzophenones (2,4-dihydroxybenzophenone, BP-1 and 2,2'-dihydroxy-4-methoxybenzophenone, BP-8) in water samples is presented. Analytes were first concentrated on the
G Subba Rao et al.
Anticancer research, 33(6), 2535-2540 (2013-06-12)
Sunscreen compounds with added benefit of skin cancer prevention have both public and commercial interests. Our earlier study using the Epstein-Barr virus early antigen in vitro assay reported on skin cancer chemoprevention potential of benzophenone sunscreens. We now report the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service