Skip to Content
Merck
All Photos(1)

Documents

103160

Sigma-Aldrich

2-Hydroxybenzophenone

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOC6H4COC6H5
CAS Number:
Molecular Weight:
198.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

bp

171-173 °C/12 mmHg (lit.)

mp

37-39 °C (lit.)

SMILES string

Oc1ccccc1C(=O)c2ccccc2

InChI

1S/C13H10O2/c14-12-9-5-4-8-11(12)13(15)10-6-2-1-3-7-10/h1-9,14H

InChI key

HJIAMFHSAAEUKR-UHFFFAOYSA-N

General description

  • 2-Hydroxybenzophenone forms copolymer 2-hydroxy, 3-allyl, 4,4′-dimethoxybenzophenone with methyl methacrylate.
  • It forms photostabilizer compounds with 2,2,6,6-tetramethylpiperidine under phase transfer catalysis conditions.
  • It reacts with 3-aminophenyl boronic acid to form macrocyclic boronates.

Application

2-Hydroxybenzophenone was used to quantify the metabolites of unmethylated extracts from the supernatants of Pseudomonas acidovorans cultures grown on 1,1-diphenylethylene.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Vanessa Giannetti et al.
Journal of the science of food and agriculture, 97(7), 2191-2198 (2016-11-02)
The lack of harmonized European legislation on food packaging led the Confederation of European Paper Industries to the proposal of a voluntary Industry Guideline for the compliance of paper and board materials for food contact applications. In the present work
Transient absorption spectra of 2-hydroxybenzophenone photostabilizers
Merritt C, et al.
Chemical Physics Letters, 69(1), 169-173 (1980)
New boron macrocycles based on self-assembly of Schiff bases
Barba V, et al.
Journal of Organometallic Chemistry, 622(1), 259-264 (2001)
Synthesis and application of new combined 2, 2, 6, 6-tetramethylpiperidine-2-hydroxybenzophenone 1, 3, 5-triazine derivatives as photostabilizers for polymer materials
Bojinov V and Grabchev I
J. Photochem. Photobiol., A, 146(3), 199-205 (2002)
A G Hay et al.
Applied and environmental microbiology, 64(6), 2141-2146 (1998-06-03)
1,1-Dichloro-2,2-bis(4-chlorophenyl)ethylene (DDE), a toxic breakdown product of 1,1,1-trichloro-2,2-bis(4-chlorophenyl)ethane (DDT), has traditionally been viewed as a dead-end metabolite: there are no published reports detailing enzymatic ring fission of DDE by bacteria in either soil or pure culture. In this study, we

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service